Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 212966188-212966447 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VASH2
Entrez Gene ID
79805
Gene Name
vasohibin 2
Gene Aliases
-
UniGene
Hs.96885
Gene Chromosome Location
Chr.1: 212935543-212991585 on Build GRCh38
Transcripts
NM_001136474.2 NM_001136475.2 NM_001301056.1 NM_024749.4 XM_011509987.2 XM_011509988.2 XM_011509989.2 XM_011509991.2 XM_011509994.1 XM_011509995.1 XM_017002351.1 XM_017002352.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6900133 c.238A>T p.T80S Chr.1: 212966281-212966281 on Build GRCh38
6271462 c.299G>T p.G100V Chr.1: 212966342-212966342 on Build GRCh38
400953 c.257G>T p.R86L Chr.1: 212966300-212966300 on Build GRCh38
5379602 c.279C>T p.C93C Chr.1: 212966322-212966322 on Build GRCh38
7403147 c.302+3A>G p.? Chr.1: 212966348-212966348 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs564740694 Chr.1: 212966398-212966398 on Build GRCh38
rs753751653 Chr.1: 212966384-212966384 on Build GRCh38
rs550520981 Chr.1: 212966407-212966407 on Build GRCh38
rs892660129 Chr.1: 212966213-212966213 on Build GRCh38
rs3738805 Chr.1: 212966406-212966406 on Build GRCh38
rs368363433 Chr.1: 212966218-212966218 on Build GRCh38
rs558983637 Chr.1: 212966300-212966300 on Build GRCh38
rs57434622 Chr.1: 212966219-212966219 on Build GRCh38
rs935491319 Chr.1: 212966406-212966407 on Build GRCh38
rs116102108 Chr.1: 212966382-212966382 on Build GRCh38
rs140154675 Chr.1: 212966229-212966229 on Build GRCh38
rs552492497 Chr.1: 212966388-212966388 on Build GRCh38
rs960694350 Chr.1: 212966389-212966389 on Build GRCh38
rs866220776 Chr.1: 212966376-212966376 on Build GRCh38
rs919315199 Chr.1: 212966404-212966404 on Build GRCh38
rs763490312 Chr.1: 212966327-212966327 on Build GRCh38
rs1035069719 Chr.1: 212966268-212966268 on Build GRCh38
rs960671397 Chr.1: 212966282-212966282 on Build GRCh38
rs890893541 Chr.1: 212966418-212966418 on Build GRCh38
rs142050670 Chr.1: 212966400-212966400 on Build GRCh38
rs775923575 Chr.1: 212966305-212966305 on Build GRCh38
rs746163095 Chr.1: 212966247-212966247 on Build GRCh38
rs991238628 Chr.1: 212966380-212966380 on Build GRCh38
rs946980370 Chr.1: 212966392-212966392 on Build GRCh38
rs764547309 Chr.1: 212966367-212966367 on Build GRCh38
rs111850024 Chr.1: 212966215-212966215 on Build GRCh38

View Full Product Details