Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 84905985-84906256 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMSB10
Entrez Gene ID
9168
Gene Name
thymosin beta 10
Gene Aliases
MIG12, TB10
UniGene
Hs.446574
Gene Chromosome Location
Chr.2: 84905639-84906675 on Build GRCh38
Transcripts
NM_021103.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4734791 c.17A>G p.D6G Chr.2: 84906034-84906034 on Build GRCh38
4001766 c.75G>A p.E25E Chr.2: 84906092-84906092 on Build GRCh38
1023315 c.66G>A p.E22E Chr.2: 84906083-84906083 on Build GRCh38
5960729 c.4G>T p.A2S Chr.2: 84906021-84906021 on Build GRCh38
1229833 c.57G>T p.K19N Chr.2: 84906074-84906074 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757649978 Chr.2: 84906043-84906043 on Build GRCh38
rs1032052720 Chr.2: 84906156-84906156 on Build GRCh38
rs781715978 Chr.2: 84906048-84906048 on Build GRCh38
rs377588100 Chr.2: 84906200-84906200 on Build GRCh38
rs1804517 Chr.2: 84906054-84906054 on Build GRCh38
rs761483424 Chr.2: 84906117-84906117 on Build GRCh38
rs148820734 Chr.2: 84906116-84906116 on Build GRCh38
rs12639 Chr.2: 84906071-84906071 on Build GRCh38
rs766759116 Chr.2: 84906217-84906217 on Build GRCh38
rs1017988669 Chr.2: 84906177-84906177 on Build GRCh38
rs986433524 Chr.2: 84906123-84906123 on Build GRCh38
rs542288958 Chr.2: 84906195-84906195 on Build GRCh38
rs751794235 Chr.2: 84906168-84906168 on Build GRCh38
rs766290420 Chr.2: 84906152-84906152 on Build GRCh38
rs546495087 Chr.2: 84906208-84906208 on Build GRCh38
rs758647259 Chr.2: 84906021-84906021 on Build GRCh38
rs199916896 Chr.2: 84906132-84906132 on Build GRCh38
rs915524228 Chr.2: 84906230-84906230 on Build GRCh38
rs556341812 Chr.2: 84906155-84906155 on Build GRCh38
rs751847702 Chr.2: 84906036-84906036 on Build GRCh38
rs878926651 Chr.2: 84906122-84906122 on Build GRCh38
rs879253064 Chr.2: 84906125-84906125 on Build GRCh38
rs773037269 Chr.2: 84906133-84906133 on Build GRCh38
rs1804515 Chr.2: 84906037-84906037 on Build GRCh38
rs564727129 Chr.2: 84906213-84906213 on Build GRCh38
rs879060281 Chr.2: 84906119-84906119 on Build GRCh38
rs878979260 Chr.2: 84906190-84906190 on Build GRCh38
rs752773544 Chr.2: 84906006-84906006 on Build GRCh38
rs934305319 Chr.2: 84906182-84906182 on Build GRCh38
rs760644166 Chr.2: 84906139-84906139 on Build GRCh38
rs576529499 Chr.2: 84906215-84906215 on Build GRCh38
rs780617613 Chr.2: 84906152-84906152 on Build GRCh38
rs878999190 Chr.2: 84906120-84906120 on Build GRCh38
rs999281006 Chr.2: 84906138-84906138 on Build GRCh38
rs764294846 Chr.2: 84906167-84906167 on Build GRCh38
rs746356857 Chr.2: 84906052-84906052 on Build GRCh38
rs771821040 Chr.2: 84906118-84906118 on Build GRCh38
rs12471929 Chr.2: 84906196-84906196 on Build GRCh38
rs1046274430 Chr.2: 84906107-84906107 on Build GRCh38
rs1055725329 Chr.2: 84906229-84906229 on Build GRCh38
rs11553756 Chr.2: 84906088-84906088 on Build GRCh38
rs779603945 Chr.2: 84906069-84906069 on Build GRCh38
rs924298858 Chr.2: 84906159-84906159 on Build GRCh38
rs140658686 Chr.2: 84906194-84906194 on Build GRCh38
rs574777968 Chr.2: 84906163-84906163 on Build GRCh38
rs7148 Chr.2: 84906092-84906092 on Build GRCh38
rs778077940 Chr.2: 84906032-84906032 on Build GRCh38
rs772403849 Chr.2: 84906142-84906142 on Build GRCh38
rs878893348 Chr.2: 84906197-84906197 on Build GRCh38
rs770253927 Chr.2: 84906068-84906068 on Build GRCh38
rs1029060593 Chr.2: 84906198-84906198 on Build GRCh38

View Full Product Details