Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 203007739-203007964 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM183A
Entrez Gene ID
92703
Gene Name
transmembrane protein 183A
Gene Aliases
C1orf37
UniGene
Hs.497443
Gene Chromosome Location
Chr.1: 203007367-203024848 on Build GRCh38
Transcripts
NM_001322955.1 NM_001322956.1 NM_001322957.1 NM_001322958.1 NM_001322959.1 NM_138391.4 NR_136530.1 NR_136531.1 NR_136532.1 NR_136533.1 NR_136534.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6989766 c.134C>T p.S45L Chr.1: 203007798-203007798 on Build GRCh38
7240044 c.114C>G p.T38T Chr.1: 203007778-203007778 on Build GRCh38
4027127 c.138T>C p.D46D Chr.1: 203007802-203007802 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs746989938 Chr.1: 203007839-203007839 on Build GRCh38
rs771860424 Chr.1: 203007826-203007826 on Build GRCh38
rs144664475 Chr.1: 203007816-203007816 on Build GRCh38
rs777872549 Chr.1: 203007838-203007838 on Build GRCh38
rs753359733 Chr.1: 203007892-203007892 on Build GRCh38
rs757638527 Chr.1: 203007782-203007782 on Build GRCh38
rs747975881 Chr.1: 203007824-203007824 on Build GRCh38
rs546483494 Chr.1: 203007873-203007873 on Build GRCh38
rs971011069 Chr.1: 203007922-203007922 on Build GRCh38
rs752383079 Chr.1: 203007909-203007909 on Build GRCh38
rs190392437 Chr.1: 203007915-203007915 on Build GRCh38
rs778655666 Chr.1: 203007906-203007906 on Build GRCh38
rs768100958 Chr.1: 203007785-203007785 on Build GRCh38
rs763030565 Chr.1: 203007848-203007848 on Build GRCh38
rs769992221 Chr.1: 203007843-203007843 on Build GRCh38
rs751971162 Chr.1: 203007778-203007778 on Build GRCh38
rs758188447 Chr.1: 203007910-203007910 on Build GRCh38
rs965826790 Chr.1: 203007811-203007811 on Build GRCh38
rs201795085 Chr.1: 203007885-203007885 on Build GRCh38
rs778514078 Chr.1: 203007823-203007823 on Build GRCh38
rs768931026 Chr.1: 203007851-203007851 on Build GRCh38
rs1008693717 Chr.1: 203007798-203007798 on Build GRCh38
rs777780297 Chr.1: 203007912-203007912 on Build GRCh38
rs754516203 Chr.1: 203007897-203007897 on Build GRCh38
rs774539984 Chr.1: 203007854-203007854 on Build GRCh38
rs375647100 Chr.1: 203007805-203007805 on Build GRCh38
rs764357364 Chr.1: 203007759-203007759 on Build GRCh38
rs767920218 Chr.1: 203007876-203007876 on Build GRCh38
rs200150391 Chr.1: 203007820-203007820 on Build GRCh38
rs1027882397 Chr.1: 203007837-203007837 on Build GRCh38
rs958582534 Chr.1: 203007845-203007845 on Build GRCh38
rs775773586 Chr.1: 203007847-203007847 on Build GRCh38
rs551330258 Chr.1: 203007796-203007796 on Build GRCh38
rs953176917 Chr.1: 203007767-203007767 on Build GRCh38
rs749526051 Chr.1: 203007902-203007902 on Build GRCh38
rs780276061 Chr.1: 203007886-203007886 on Build GRCh38
rs140775257 Chr.1: 203007806-203007806 on Build GRCh38
rs566698212 Chr.1: 203007879-203007879 on Build GRCh38

View Full Product Details