Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 170945458-170945683 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MROH9
Entrez Gene ID
80133
Gene Name
maestro heat like repeat family member 9
Gene Aliases
ARMC11, C1orf129
UniGene
Hs.591489
Gene Chromosome Location
Chr.1: 170935471-171066341 on Build GRCh38
Transcripts
NM_001163629.1 NM_025063.2 XM_011510005.2 XM_011510006.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3478199 c.11G>A p.R4K Chr.1: 170945567-170945567 on Build GRCh38
6527881 c.12G>C p.R4S Chr.1: 170945568-170945568 on Build GRCh38
6437606 c.14A>G p.N5S Chr.1: 170945570-170945570 on Build GRCh38
463406 c.17C>T p.P6L Chr.1: 170945573-170945573 on Build GRCh38
243328 c.9A>G p.T3T Chr.1: 170945565-170945565 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752103626 Chr.1: 170945511-170945511 on Build GRCh38
rs892733819 Chr.1: 170945636-170945636 on Build GRCh38
rs778733713 Chr.1: 170945560-170945560 on Build GRCh38
rs149089755 Chr.1: 170945597-170945597 on Build GRCh38
rs770775310 Chr.1: 170945601-170945601 on Build GRCh38
rs758093815 Chr.1: 170945537-170945537 on Build GRCh38
rs923349600 Chr.1: 170945527-170945527 on Build GRCh38
rs754641089 Chr.1: 170945546-170945546 on Build GRCh38
rs949099413 Chr.1: 170945611-170945611 on Build GRCh38
rs1001021524 Chr.1: 170945628-170945628 on Build GRCh38
rs759633503 Chr.1: 170945616-170945616 on Build GRCh38
rs745480875 Chr.1: 170945509-170945509 on Build GRCh38
rs371508287 Chr.1: 170945537-170945537 on Build GRCh38
rs758768009 Chr.1: 170945508-170945508 on Build GRCh38
rs747856311 Chr.1: 170945571-170945571 on Build GRCh38
rs764406295 Chr.1: 170945509-170945509 on Build GRCh38
rs1009222205 Chr.1: 170945638-170945638 on Build GRCh38
rs779561113 Chr.1: 170945536-170945536 on Build GRCh38
rs771873227 Chr.1: 170945577-170945577 on Build GRCh38
rs746799519 Chr.1: 170945529-170945529 on Build GRCh38
rs889289100 Chr.1: 170945618-170945619 on Build GRCh38
rs1053960593 Chr.1: 170945631-170945632 on Build GRCh38
rs1025080430 Chr.1: 170945640-170945640 on Build GRCh38
rs757763007 Chr.1: 170945528-170945528 on Build GRCh38
rs769910039 Chr.1: 170945626-170945626 on Build GRCh38
rs775657720 Chr.1: 170945631-170945631 on Build GRCh38
rs376152654 Chr.1: 170945596-170945596 on Build GRCh38

View Full Product Details