Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6453184-6453425 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAPBPL
Entrez Gene ID
55080
Gene Name
TAP binding protein like
Gene Aliases
TAPBP-R, TAPBPR
UniGene
Hs.504597
Gene Chromosome Location
Chr.12: 6451655-6472006 on Build GRCh38
Transcripts
NM_018009.4 XM_005253700.3 XM_017019545.1 XM_017019546.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6419501 c.191T>C p.L64P Chr.12: 6453193-6453193 on Build GRCh38
5639864 c.245A>G p.Q82R Chr.12: 6453247-6453247 on Build GRCh38
1744516 c.290C>A p.A97D Chr.12: 6453292-6453292 on Build GRCh38
4412296 c.199G>T p.V67L Chr.12: 6453201-6453201 on Build GRCh38
942555 c.288G>T p.E96D Chr.12: 6453290-6453290 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774225029 Chr.12: 6453401-6453401 on Build GRCh38
rs750096943 Chr.12: 6453223-6453223 on Build GRCh38
rs578155041 Chr.12: 6453248-6453248 on Build GRCh38
rs752494976 Chr.12: 6453278-6453278 on Build GRCh38
rs778624801 Chr.12: 6453215-6453215 on Build GRCh38
rs2286722 Chr.12: 6453310-6453310 on Build GRCh38
rs756174566 Chr.12: 6453286-6453286 on Build GRCh38
rs189877599 Chr.12: 6453363-6453363 on Build GRCh38
rs766147694 Chr.12: 6453228-6453228 on Build GRCh38
rs759616853 Chr.12: 6453262-6453262 on Build GRCh38
rs376284563 Chr.12: 6453332-6453332 on Build GRCh38
rs781635453 Chr.12: 6453322-6453322 on Build GRCh38
rs540000792 Chr.12: 6453282-6453282 on Build GRCh38
rs896208233 Chr.12: 6453368-6453368 on Build GRCh38
rs753465657 Chr.12: 6453293-6453293 on Build GRCh38
rs750236251 Chr.12: 6453313-6453318 on Build GRCh38
rs986377714 Chr.12: 6453273-6453273 on Build GRCh38
rs747011459 Chr.12: 6453334-6453334 on Build GRCh38
rs2041386 Chr.12: 6453372-6453372 on Build GRCh38
rs756779168 Chr.12: 6453298-6453298 on Build GRCh38
rs771582560 Chr.12: 6453320-6453320 on Build GRCh38
rs777825441 Chr.12: 6453292-6453292 on Build GRCh38
rs751789179 Chr.12: 6453259-6453259 on Build GRCh38
rs745541127 Chr.12: 6453315-6453315 on Build GRCh38
rs1044743234 Chr.12: 6453378-6453378 on Build GRCh38
rs766573134 Chr.12: 6453253-6453253 on Build GRCh38
rs767346246 Chr.12: 6453270-6453270 on Build GRCh38
rs182450186 Chr.12: 6453388-6453388 on Build GRCh38
rs749423872 Chr.12: 6453341-6453341 on Build GRCh38
rs553972468 Chr.12: 6453327-6453327 on Build GRCh38
rs146581116 Chr.12: 6453216-6453216 on Build GRCh38
rs771053633 Chr.12: 6453397-6453397 on Build GRCh38
rs770751415 Chr.12: 6453209-6453209 on Build GRCh38
rs569968024 Chr.12: 6453271-6453271 on Build GRCh38
rs770234991 Chr.12: 6453235-6453235 on Build GRCh38
rs762338228 Chr.12: 6453233-6453233 on Build GRCh38
rs758069754 Chr.12: 6453326-6453326 on Build GRCh38
rs995172572 Chr.12: 6453389-6453389 on Build GRCh38
rs371473125 Chr.12: 6453336-6453336 on Build GRCh38
rs773328258 Chr.12: 6453238-6453238 on Build GRCh38
rs141168269 Chr.12: 6453240-6453240 on Build GRCh38
rs199504335 Chr.12: 6453331-6453331 on Build GRCh38

View Full Product Details