Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 114168151-114168382 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL27A1
Entrez Gene ID
85301
Gene Name
collagen type XXVII alpha 1
Gene Aliases
STLS
UniGene
Hs.494892
Gene Chromosome Location
Chr.9: 114154271-114312516 on Build GRCh38
Transcripts
NM_032888.3 XM_006717308.2 XM_011519138.2 XM_011519140.1 XM_011519142.2 XM_011519143.2 XM_011519144.2 XM_017015239.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4837895 c.718G>A p.D240N Chr.9: 114168273-114168273 on Build GRCh38
3903348 c.750C>A p.F250L Chr.9: 114168305-114168305 on Build GRCh38
5359660 c.612G>A p.P204P Chr.9: 114168167-114168167 on Build GRCh38
3903347 c.673G>A p.A225T Chr.9: 114168228-114168228 on Build GRCh38
3847436 c.808G>A p.E270K Chr.9: 114168363-114168363 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199729771 Chr.9: 114168205-114168205 on Build GRCh38
rs574675570 Chr.9: 114168278-114168278 on Build GRCh38
rs200344636 Chr.9: 114168342-114168342 on Build GRCh38
rs918738464 Chr.9: 114168311-114168311 on Build GRCh38
rs771709770 Chr.9: 114168228-114168228 on Build GRCh38
rs745911804 Chr.9: 114168221-114168221 on Build GRCh38
rs867317084 Chr.9: 114168279-114168279 on Build GRCh38
rs749690330 Chr.9: 114168330-114168330 on Build GRCh38
rs756760165 Chr.9: 114168224-114168224 on Build GRCh38
rs146040174 Chr.9: 114168325-114168325 on Build GRCh38
rs148242800 Chr.9: 114168340-114168340 on Build GRCh38
rs369005326 Chr.9: 114168233-114168233 on Build GRCh38
rs376160858 Chr.9: 114168315-114168315 on Build GRCh38
rs374669935 Chr.9: 114168227-114168227 on Build GRCh38
rs778225067 Chr.9: 114168329-114168329 on Build GRCh38
rs538713049 Chr.9: 114168236-114168236 on Build GRCh38
rs34578955 Chr.9: 114168348-114168348 on Build GRCh38
rs938788154 Chr.9: 114168295-114168295 on Build GRCh38
rs759744480 Chr.9: 114168203-114168203 on Build GRCh38
rs778424647 Chr.9: 114168225-114168225 on Build GRCh38
rs1037173893 Chr.9: 114168298-114168298 on Build GRCh38
rs546860616 Chr.9: 114168257-114168257 on Build GRCh38
rs764318865 Chr.9: 114168354-114168354 on Build GRCh38
rs767653538 Chr.9: 114168204-114168204 on Build GRCh38
rs980081961 Chr.9: 114168292-114168292 on Build GRCh38
rs1030771876 Chr.9: 114168349-114168349 on Build GRCh38
rs757891945 Chr.9: 114168297-114168297 on Build GRCh38
rs757163848 Chr.9: 114168287-114168287 on Build GRCh38
rs148448277 Chr.9: 114168277-114168277 on Build GRCh38
rs751595585 Chr.9: 114168198-114168198 on Build GRCh38
rs144556713 Chr.9: 114168268-114168268 on Build GRCh38
rs970026412 Chr.9: 114168281-114168281 on Build GRCh38
rs144068722 Chr.9: 114168294-114168294 on Build GRCh38
rs763107645 Chr.9: 114168189-114168189 on Build GRCh38
rs557055064 Chr.9: 114168347-114168347 on Build GRCh38
rs369411223 Chr.9: 114168355-114168355 on Build GRCh38
rs753638362 Chr.9: 114168284-114168284 on Build GRCh38
rs144086565 Chr.9: 114168316-114168316 on Build GRCh38
rs773900818 Chr.9: 114168185-114168185 on Build GRCh38
rs201345287 Chr.9: 114168341-114168341 on Build GRCh38
rs779610682 Chr.9: 114168231-114168231 on Build GRCh38
rs201192948 Chr.9: 114168248-114168248 on Build GRCh38
rs772101697 Chr.9: 114168345-114168345 on Build GRCh38
rs372325271 Chr.9: 114168220-114168220 on Build GRCh38
rs766546940 Chr.9: 114168191-114168191 on Build GRCh38
rs143912297 Chr.9: 114168174-114168174 on Build GRCh38
rs149260799 Chr.9: 114168263-114168263 on Build GRCh38
rs773847766 Chr.9: 114168238-114168238 on Build GRCh38
rs895361431 Chr.9: 114168332-114168332 on Build GRCh38
rs912387390 Chr.9: 114168254-114168254 on Build GRCh38
rs778741093 Chr.9: 114168288-114168288 on Build GRCh38
rs763485848 Chr.9: 114168244-114168244 on Build GRCh38

View Full Product Details