Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 113205988-113206239 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCAM1
Entrez Gene ID
4684
Gene Name
neural cell adhesion molecule 1
Gene Aliases
CD56, MSK39, NCAM
UniGene
Hs.503878
Gene Chromosome Location
Chr.11: 112961247-113278436 on Build GRCh38
Transcripts
NM_000615.6 NM_001076682.3 NM_001242607.1 NM_001242608.1 NM_181351.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4902868 c.570C>A p.G190G Chr.11: 113206125-113206125 on Build GRCh38
428423 c.547G>A p.E183K Chr.11: 113206102-113206102 on Build GRCh38
3443435 c.562G>A p.E188K Chr.11: 113206144-113206144 on Build GRCh38
1475042 c.525C>T p.I175I Chr.11: 113206080-113206080 on Build GRCh38
5535236 c.615C>T p.V205V Chr.11: 113206170-113206170 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781929864 Chr.11: 113206188-113206188 on Build GRCh38
rs782535732 Chr.11: 113206091-113206091 on Build GRCh38
rs377417619 Chr.11: 113206194-113206194 on Build GRCh38
rs782517960 Chr.11: 113206203-113206203 on Build GRCh38
rs782014259 Chr.11: 113206146-113206146 on Build GRCh38
rs782242910 Chr.11: 113206136-113206136 on Build GRCh38
rs35576001 Chr.11: 113206082-113206082 on Build GRCh38
rs576548045 Chr.11: 113206138-113206138 on Build GRCh38
rs556599914 Chr.11: 113206097-113206097 on Build GRCh38
rs782807183 Chr.11: 113206016-113206016 on Build GRCh38
rs949627474 Chr.11: 113206156-113206156 on Build GRCh38
rs782184114 Chr.11: 113206105-113206105 on Build GRCh38
rs376708434 Chr.11: 113206115-113206115 on Build GRCh38
rs781819663 Chr.11: 113206043-113206043 on Build GRCh38
rs369920955 Chr.11: 113206193-113206193 on Build GRCh38
rs559320851 Chr.11: 113206200-113206200 on Build GRCh38
rs782683789 Chr.11: 113206183-113206183 on Build GRCh38
rs782161510 Chr.11: 113206165-113206165 on Build GRCh38
rs782579837 Chr.11: 113206039-113206039 on Build GRCh38
rs782494134 Chr.11: 113206056-113206056 on Build GRCh38
rs782329784 Chr.11: 113206109-113206109 on Build GRCh38
rs782813348 Chr.11: 113206206-113206206 on Build GRCh38
rs782525915 Chr.11: 113206022-113206022 on Build GRCh38
rs782632084 Chr.11: 113206081-113206081 on Build GRCh38

View Full Product Details