Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 60503734-60503991 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MS4A12
Entrez Gene ID
54860
Gene Name
membrane spanning 4-domains A12
Gene Aliases
Ms4a10
UniGene
Hs.272789
Gene Chromosome Location
Chr.11: 60492743-60507430 on Build GRCh38
Transcripts
NM_001164470.1 NM_017716.2 XM_011545117.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5434264 c.524G>C p.G175A Chr.11: 60503753-60503753 on Build GRCh38
2036329 c.547A>C p.M183L Chr.11: 60503776-60503776 on Build GRCh38
689409 c.559G>C p.G187R Chr.11: 60503788-60503788 on Build GRCh38
4703293 c.584C>T p.A195V Chr.11: 60503813-60503813 on Build GRCh38
6045606 c.572A>G p.Q191R Chr.11: 60503801-60503801 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs969977860 Chr.11: 60503894-60503894 on Build GRCh38
rs757007159 Chr.11: 60503842-60503842 on Build GRCh38
rs773669521 Chr.11: 60503774-60503774 on Build GRCh38
rs562706044 Chr.11: 60503863-60503863 on Build GRCh38
rs2298557 Chr.11: 60503864-60503864 on Build GRCh38
rs760157189 Chr.11: 60503758-60503758 on Build GRCh38
rs909430282 Chr.11: 60503942-60503942 on Build GRCh38
rs868107876 Chr.11: 60503897-60503897 on Build GRCh38
rs887554023 Chr.11: 60503846-60503846 on Build GRCh38
rs766101531 Chr.11: 60503911-60503911 on Build GRCh38
rs774823969 Chr.11: 60503869-60503869 on Build GRCh38
rs771226235 Chr.11: 60503814-60503814 on Build GRCh38
rs745636724 Chr.11: 60503848-60503848 on Build GRCh38
rs755297132 Chr.11: 60503810-60503810 on Build GRCh38
rs751736928 Chr.11: 60503801-60503801 on Build GRCh38
rs374195006 Chr.11: 60503770-60503770 on Build GRCh38
rs778833410 Chr.11: 60503847-60503847 on Build GRCh38
rs760389379 Chr.11: 60503872-60503872 on Build GRCh38
rs368371684 Chr.11: 60503824-60503824 on Build GRCh38
rs35906967 Chr.11: 60503860-60503861 on Build GRCh38
rs116528387 Chr.11: 60503786-60503786 on Build GRCh38
rs780542886 Chr.11: 60503800-60503800 on Build GRCh38
rs767671907 Chr.11: 60503867-60503871 on Build GRCh38
rs768106321 Chr.11: 60503859-60503859 on Build GRCh38
rs767822239 Chr.11: 60503815-60503815 on Build GRCh38
rs371494653 Chr.11: 60503826-60503826 on Build GRCh38
rs544946315 Chr.11: 60503952-60503952 on Build GRCh38
rs201039425 Chr.11: 60503829-60503829 on Build GRCh38
rs983485776 Chr.11: 60503906-60503906 on Build GRCh38
rs577537930 Chr.11: 60503776-60503776 on Build GRCh38
rs773726398 Chr.11: 60503860-60503860 on Build GRCh38
rs2298556 Chr.11: 60503852-60503852 on Build GRCh38
rs760762471 Chr.11: 60503895-60503895 on Build GRCh38
rs748529534 Chr.11: 60503840-60503840 on Build GRCh38
rs965219984 Chr.11: 60503856-60503856 on Build GRCh38

View Full Product Details