Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 70732841-70733072 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP3K9
Entrez Gene ID
4293
Gene Name
mitogen-activated protein kinase kinase kinase 9
Gene Aliases
MEKK9, MLK1, PRKE1
UniGene
Hs.445496
Gene Chromosome Location
Chr.14: 70722526-70809268 on Build GRCh38
Transcripts
NM_001284230.1 NM_001284231.1 NM_001284232.1 NM_033141.3 XM_005267683.4 XM_011536788.2 XM_011536792.1 XM_011536794.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
957421 c.2565C>T p.S855S Chr.14: 70732846-70732846 on Build GRCh38
957422 c.2421A>G p.R807R Chr.14: 70732990-70732990 on Build GRCh38
5616787 c.2493G>A p.K831K Chr.14: 70732918-70732918 on Build GRCh38
4819111 c.2564C>G p.S855C Chr.14: 70732847-70732847 on Build GRCh38
1239399 c.2458C>T p.R820W Chr.14: 70732953-70732953 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779504029 Chr.14: 70732881-70732881 on Build GRCh38
rs759804164 Chr.14: 70733038-70733038 on Build GRCh38
rs755998074 Chr.14: 70732999-70733001 on Build GRCh38
rs766443716 Chr.14: 70733025-70733025 on Build GRCh38
rs991014475 Chr.14: 70732925-70732925 on Build GRCh38
rs779790828 Chr.14: 70733004-70733004 on Build GRCh38
rs375769231 Chr.14: 70732931-70732931 on Build GRCh38
rs752591387 Chr.14: 70732936-70732937 on Build GRCh38
rs940301306 Chr.14: 70732979-70732979 on Build GRCh38
rs949120910 Chr.14: 70732956-70732956 on Build GRCh38
rs780975980 Chr.14: 70732901-70732901 on Build GRCh38
rs201326431 Chr.14: 70733022-70733022 on Build GRCh38
rs545709185 Chr.14: 70732902-70732902 on Build GRCh38
rs376367504 Chr.14: 70732937-70732937 on Build GRCh38
rs572178360 Chr.14: 70732870-70732870 on Build GRCh38
rs755389971 Chr.14: 70732871-70732871 on Build GRCh38
rs745488869 Chr.14: 70732988-70732988 on Build GRCh38
rs778571437 Chr.14: 70733039-70733039 on Build GRCh38
rs769286914 Chr.14: 70732989-70732989 on Build GRCh38
rs746450605 Chr.14: 70732887-70732887 on Build GRCh38
rs367998651 Chr.14: 70733009-70733009 on Build GRCh38
rs1014191193 Chr.14: 70733045-70733045 on Build GRCh38
rs753312100 Chr.14: 70732958-70732958 on Build GRCh38
rs770575759 Chr.14: 70732895-70732895 on Build GRCh38
rs753220614 Chr.14: 70733048-70733048 on Build GRCh38
rs768440980 Chr.14: 70732924-70732924 on Build GRCh38
rs748018900 Chr.14: 70733047-70733047 on Build GRCh38
rs936340633 Chr.14: 70732898-70732898 on Build GRCh38
rs527720055 Chr.14: 70732962-70732962 on Build GRCh38
rs776922633 Chr.14: 70733033-70733033 on Build GRCh38
rs755374301 Chr.14: 70732955-70732955 on Build GRCh38
rs761825090 Chr.14: 70732932-70732932 on Build GRCh38
rs977982660 Chr.14: 70732885-70732885 on Build GRCh38
rs762832980 Chr.14: 70732923-70732923 on Build GRCh38
rs775202278 Chr.14: 70732920-70732920 on Build GRCh38
rs772109137 Chr.14: 70733013-70733013 on Build GRCh38
rs748151841 Chr.14: 70733010-70733010 on Build GRCh38
rs768511767 Chr.14: 70733007-70733007 on Build GRCh38
rs766645408 Chr.14: 70732946-70732946 on Build GRCh38
rs368928316 Chr.14: 70733006-70733006 on Build GRCh38
rs769676505 Chr.14: 70732919-70732919 on Build GRCh38
rs756820633 Chr.14: 70732961-70732961 on Build GRCh38
rs760927645 Chr.14: 70732945-70732945 on Build GRCh38
rs754111307 Chr.14: 70732953-70732953 on Build GRCh38

View Full Product Details