Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 112996947-112997194 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTPBP8
Entrez Gene ID
29083
Gene Name
GTP binding protein 8 (putative)
Gene Aliases
HSPC135
UniGene
Hs.127496
Gene Chromosome Location
Chr.3: 112989275-113001966 on Build GRCh38
Transcripts
NM_014170.2 NM_138485.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
257075 c.646G>T p.E216* Chr.3: 112996971-112996971 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1043990810 Chr.3: 112997152-112997152 on Build GRCh38
rs769532104 Chr.3: 112997002-112997002 on Build GRCh38
rs370330282 Chr.3: 112997015-112997015 on Build GRCh38
rs971782535 Chr.3: 112997021-112997021 on Build GRCh38
rs913932523 Chr.3: 112997089-112997089 on Build GRCh38
rs988518601 Chr.3: 112997088-112997088 on Build GRCh38
rs747932572 Chr.3: 112997000-112997000 on Build GRCh38
rs754568290 Chr.3: 112997041-112997041 on Build GRCh38
rs763159660 Chr.3: 112996988-112996988 on Build GRCh38
rs559803137 Chr.3: 112996993-112996993 on Build GRCh38
rs374745070 Chr.3: 112997028-112997028 on Build GRCh38
rs569952287 Chr.3: 112997072-112997072 on Build GRCh38
rs756617464 Chr.3: 112997084-112997084 on Build GRCh38
rs776484600 Chr.3: 112996978-112996978 on Build GRCh38
rs910255678 Chr.3: 112997071-112997071 on Build GRCh38
rs150743391 Chr.3: 112997123-112997123 on Build GRCh38
rs764066697 Chr.3: 112996989-112996989 on Build GRCh38
rs374400036 Chr.3: 112997000-112997000 on Build GRCh38
rs774623573 Chr.3: 112997010-112997011 on Build GRCh38
rs927998000 Chr.3: 112997049-112997049 on Build GRCh38
rs932774129 Chr.3: 112997164-112997164 on Build GRCh38
rs780777836 Chr.3: 112997031-112997031 on Build GRCh38
rs549674760 Chr.3: 112997146-112997146 on Build GRCh38
rs551553740 Chr.3: 112997045-112997045 on Build GRCh38
rs80126769 Chr.3: 112997159-112997159 on Build GRCh38
rs200408169 Chr.3: 112997040-112997040 on Build GRCh38

View Full Product Details