Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7415591-7415844 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NLGN2
Entrez Gene ID
57555
Gene Name
neuroligin 2
Gene Aliases
-
UniGene
Hs.26229
Gene Chromosome Location
Chr.17: 7405492-7419864 on Build GRCh38
Transcripts
NM_020795.3 XM_005256744.3 XM_017024897.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4989535 c.1321C>T p.R441W Chr.17: 7415794-7415794 on Build GRCh38
3742442 c.1149C>G p.N383K Chr.17: 7415622-7415622 on Build GRCh38
437366 c.1219G>C p.D407H Chr.17: 7415692-7415692 on Build GRCh38
6726703 c.1358C>T p.A453V Chr.17: 7415831-7415831 on Build GRCh38
1386276 c.1282C>T p.R428W Chr.17: 7415755-7415755 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746485119 Chr.17: 7415617-7415617 on Build GRCh38
rs1021621045 Chr.17: 7415766-7415766 on Build GRCh38
rs765555488 Chr.17: 7415727-7415727 on Build GRCh38
rs199509090 Chr.17: 7415802-7415802 on Build GRCh38
rs772822051 Chr.17: 7415670-7415670 on Build GRCh38
rs1002727127 Chr.17: 7415664-7415664 on Build GRCh38
rs924724424 Chr.17: 7415795-7415795 on Build GRCh38
rs201659845 Chr.17: 7415646-7415646 on Build GRCh38
rs767550417 Chr.17: 7415824-7415824 on Build GRCh38
rs754582240 Chr.17: 7415628-7415628 on Build GRCh38
rs749178079 Chr.17: 7415671-7415671 on Build GRCh38
rs780868011 Chr.17: 7415637-7415637 on Build GRCh38
rs774324796 Chr.17: 7415700-7415700 on Build GRCh38
rs140208582 Chr.17: 7415791-7415791 on Build GRCh38
rs929016372 Chr.17: 7415625-7415625 on Build GRCh38
rs145633225 Chr.17: 7415805-7415805 on Build GRCh38
rs144434883 Chr.17: 7415709-7415709 on Build GRCh38
rs900012397 Chr.17: 7415784-7415784 on Build GRCh38
rs747703660 Chr.17: 7415804-7415804 on Build GRCh38
rs758837565 Chr.17: 7415737-7415737 on Build GRCh38
rs777524391 Chr.17: 7415806-7415806 on Build GRCh38
rs754813724 Chr.17: 7415619-7415619 on Build GRCh38
rs763186967 Chr.17: 7415613-7415613 on Build GRCh38
rs770829525 Chr.17: 7415685-7415685 on Build GRCh38
rs750746672 Chr.17: 7415736-7415736 on Build GRCh38
rs376668595 Chr.17: 7415726-7415726 on Build GRCh38
rs770668096 Chr.17: 7415823-7415823 on Build GRCh38
rs564902048 Chr.17: 7415716-7415716 on Build GRCh38
rs12947017 Chr.17: 7415616-7415616 on Build GRCh38
rs766969799 Chr.17: 7415746-7415746 on Build GRCh38
rs769488635 Chr.17: 7415661-7415661 on Build GRCh38
rs747203547 Chr.17: 7415643-7415643 on Build GRCh38
rs752059943 Chr.17: 7415747-7415747 on Build GRCh38
rs968765404 Chr.17: 7415790-7415790 on Build GRCh38
rs749019100 Chr.17: 7415814-7415814 on Build GRCh38
rs958677713 Chr.17: 7415686-7415686 on Build GRCh38

View Full Product Details