Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 63912362-63912593 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VPS54
Entrez Gene ID
51542
Gene Name
VPS54, GARP complex subunit
Gene Aliases
HCC8, PPP1R164, SLP-8p, VPS54L, WR, hVps54L
UniGene
Hs.48499
Gene Chromosome Location
Chr.2: 63892149-64019421 on Build GRCh38
Transcripts
NM_001005739.1 NM_016516.2 XM_006712029.3 XM_017004316.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4095117 c.2582T>A p.L861H Chr.2: 63912388-63912388 on Build GRCh38
4740586 c.2554G>T p.D852Y Chr.2: 63912416-63912416 on Build GRCh38
1408973 c.2537T>C p.I846T Chr.2: 63912547-63912547 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs571291727 Chr.2: 63912513-63912513 on Build GRCh38
rs754812683 Chr.2: 63912529-63912529 on Build GRCh38
rs747705677 Chr.2: 63912475-63912475 on Build GRCh38
rs780029607 Chr.2: 63912448-63912448 on Build GRCh38
rs755953054 Chr.2: 63912520-63912520 on Build GRCh38
rs375996471 Chr.2: 63912447-63912447 on Build GRCh38
rs749221735 Chr.2: 63912454-63912454 on Build GRCh38
rs771332326 Chr.2: 63912504-63912504 on Build GRCh38
rs751889972 Chr.2: 63912519-63912519 on Build GRCh38
rs778236377 Chr.2: 63912464-63912464 on Build GRCh38
rs768667320 Chr.2: 63912458-63912458 on Build GRCh38
rs764407618 Chr.2: 63912517-63912517 on Build GRCh38
rs372665741 Chr.2: 63912512-63912512 on Build GRCh38
rs182165174 Chr.2: 63912524-63912524 on Build GRCh38
rs1006457723 Chr.2: 63912551-63912551 on Build GRCh38
rs139787198 Chr.2: 63912496-63912496 on Build GRCh38
rs765277014 Chr.2: 63912397-63912397 on Build GRCh38
rs771641438 Chr.2: 63912494-63912494 on Build GRCh38
rs777167594 Chr.2: 63912505-63912505 on Build GRCh38
rs758649070 Chr.2: 63912435-63912435 on Build GRCh38
rs778913454 Chr.2: 63912561-63912561 on Build GRCh38
rs755059960 Chr.2: 63912394-63912394 on Build GRCh38
rs745591184 Chr.2: 63912442-63912442 on Build GRCh38
rs779672475 Chr.2: 63912523-63912523 on Build GRCh38
rs901607668 Chr.2: 63912480-63912480 on Build GRCh38
rs149005051 Chr.2: 63912419-63912419 on Build GRCh38
rs753687856 Chr.2: 63912527-63912527 on Build GRCh38
rs149654945 Chr.2: 63912418-63912418 on Build GRCh38
rs759923569 Chr.2: 63912507-63912507 on Build GRCh38
rs372695754 Chr.2: 63912440-63912440 on Build GRCh38
rs772910747 Chr.2: 63912495-63912495 on Build GRCh38
rs17857260 Chr.2: 63912553-63912553 on Build GRCh38
rs772285527 Chr.2: 63912531-63912531 on Build GRCh38
rs112476657 Chr.2: 63912497-63912497 on Build GRCh38
rs372263623 Chr.2: 63912521-63912521 on Build GRCh38
rs746426673 Chr.2: 63912525-63912525 on Build GRCh38

View Full Product Details