Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 154551776-154552009 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBE2Q1
Entrez Gene ID
55585
Gene Name
ubiquitin conjugating enzyme E2 Q1
Gene Aliases
GTAP, NICE-5, PRO3094, UBE2Q
UniGene
Hs.607928
Gene Chromosome Location
Chr.1: 154548575-154558644 on Build GRCh38
Transcripts
NM_017582.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3802031 c.1038C>T p.G346G Chr.1: 154551807-154551807 on Build GRCh38
6815153 c.1040G>A p.G347E Chr.1: 154551805-154551805 on Build GRCh38
7320697 c.992T>C p.F331S Chr.1: 154551954-154551954 on Build GRCh38
4790123 c.1043G>T p.G348V Chr.1: 154551802-154551802 on Build GRCh38
5820222 c.998G>T p.R333M Chr.1: 154551948-154551948 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769264431 Chr.1: 154551986-154551986 on Build GRCh38
rs201248339 Chr.1: 154551807-154551807 on Build GRCh38
rs772540313 Chr.1: 154551839-154551839 on Build GRCh38
rs752992568 Chr.1: 154551891-154551891 on Build GRCh38
rs567031061 Chr.1: 154551857-154551857 on Build GRCh38
rs777917876 Chr.1: 154551904-154551904 on Build GRCh38
rs746853939 Chr.1: 154551912-154551912 on Build GRCh38
rs780802766 Chr.1: 154551946-154551946 on Build GRCh38
rs200513754 Chr.1: 154551989-154551989 on Build GRCh38
rs763559222 Chr.1: 154551842-154551842 on Build GRCh38
rs200983573 Chr.1: 154551982-154551982 on Build GRCh38
rs762211759 Chr.1: 154551981-154551982 on Build GRCh38
rs369251905 Chr.1: 154551835-154551835 on Build GRCh38
rs753845499 Chr.1: 154551906-154551906 on Build GRCh38
rs767675858 Chr.1: 154551889-154551889 on Build GRCh38
rs569518271 Chr.1: 154551819-154551819 on Build GRCh38
rs758396146 Chr.1: 154551908-154551908 on Build GRCh38
rs373987444 Chr.1: 154551874-154551874 on Build GRCh38
rs780838823 Chr.1: 154551982-154551983 on Build GRCh38
rs768464949 Chr.1: 154551940-154551940 on Build GRCh38
rs137942282 Chr.1: 154551932-154551932 on Build GRCh38
rs757287266 Chr.1: 154551983-154551984 on Build GRCh38
rs779953144 Chr.1: 154551909-154551909 on Build GRCh38
rs376768693 Chr.1: 154551832-154551832 on Build GRCh38
rs747852566 Chr.1: 154551959-154551959 on Build GRCh38
rs377684621 Chr.1: 154551875-154551875 on Build GRCh38
rs201949684 Chr.1: 154551981-154551981 on Build GRCh38
rs756239158 Chr.1: 154551893-154551893 on Build GRCh38

View Full Product Details