Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 4662322-4662571 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLPP6
Entrez Gene ID
403313
Gene Name
phospholipid phosphatase 6
Gene Aliases
PDP1, PPAPDC2, PSDP, bA6J24.6
UniGene
Hs.107510
Gene Chromosome Location
Chr.9: 4662294-4665274 on Build GRCh38
Transcripts
NM_203453.3 NM_001039395.3 XM_006716795.2 XM_006716798.3 XM_006716799.3 XM_006716801.2 XM_011517954.1 XM_011517956.1 XM_011517957.1 XM_017014881.1 XM_017014882.1 XM_017014883.1 XM_017014885.1

Gene Symbol
SPATA6L
Entrez Gene ID
55064
Gene Name
spermatogenesis associated 6 like
Gene Aliases
C9orf68, bA6J24.2
UniGene
Hs.179615
Gene Chromosome Location
Chr.9: 4540092-4666674 on Build GRCh38
Transcripts
NM_203453.3 NM_001039395.3 XM_006716795.2 XM_006716798.3 XM_006716799.3 XM_006716801.2 XM_011517954.1 XM_011517956.1 XM_011517957.1 XM_017014881.1 XM_017014882.1 XM_017014883.1 XM_017014885.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6388772 c.134G>A p.S45N Chr.9: 4662509-4662509 on Build GRCh38
6205049 c.19A>G p.S7G Chr.9: 4662394-4662394 on Build GRCh38
3907498 c.131T>A p.L44H Chr.9: 4662506-4662506 on Build GRCh38
3907497 c.42C>T p.G14G Chr.9: 4662417-4662417 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs765775167 Chr.9: 4662367-4662367 on Build GRCh38
rs774159286 Chr.9: 4662408-4662408 on Build GRCh38
rs780790260 Chr.9: 4662385-4662385 on Build GRCh38
rs768532470 Chr.9: 4662403-4662403 on Build GRCh38
rs948663822 Chr.9: 4662355-4662355 on Build GRCh38
rs764513829 Chr.9: 4662437-4662437 on Build GRCh38
rs751985996 Chr.9: 4662446-4662446 on Build GRCh38
rs770614370 Chr.9: 4662348-4662348 on Build GRCh38
rs753306833 Chr.9: 4662549-4662549 on Build GRCh38
rs746503491 Chr.9: 4662347-4662347 on Build GRCh38
rs369535277 Chr.9: 4662477-4662477 on Build GRCh38
rs769784405 Chr.9: 4662360-4662360 on Build GRCh38
rs759899043 Chr.9: 4662366-4662366 on Build GRCh38
rs764701852 Chr.9: 4662521-4662521 on Build GRCh38
rs749206546 Chr.9: 4662480-4662480 on Build GRCh38
rs301487 Chr.9: 4662369-4662369 on Build GRCh38
rs757918538 Chr.9: 4662342-4662342 on Build GRCh38
rs756902395 Chr.9: 4662447-4662447 on Build GRCh38
rs867401574 Chr.9: 4662509-4662509 on Build GRCh38
rs548807504 Chr.9: 4662356-4662356 on Build GRCh38
rs751297429 Chr.9: 4662375-4662375 on Build GRCh38
rs956700342 Chr.9: 4662411-4662411 on Build GRCh38
rs756992431 Chr.9: 4662380-4662380 on Build GRCh38
rs1037801168 Chr.9: 4662482-4662482 on Build GRCh38
rs771140540 Chr.9: 4662494-4662494 on Build GRCh38
rs754890051 Chr.9: 4662485-4662485 on Build GRCh38
rs776898523 Chr.9: 4662498-4662498 on Build GRCh38
rs914680464 Chr.9: 4662351-4662351 on Build GRCh38
rs35791393 Chr.9: 4662398-4662398 on Build GRCh38
rs758857857 Chr.9: 4662371-4662371 on Build GRCh38
rs555147147 Chr.9: 4662502-4662502 on Build GRCh38
rs774707398 Chr.9: 4662429-4662430 on Build GRCh38
rs774851123 Chr.9: 4662522-4662522 on Build GRCh38
rs767253868 Chr.9: 4662525-4662525 on Build GRCh38
rs769958094 Chr.9: 4662504-4662504 on Build GRCh38
rs1010667552 Chr.9: 4662545-4662545 on Build GRCh38
rs767343788 Chr.9: 4662464-4662464 on Build GRCh38
rs71496487 Chr.9: 4662365-4662365 on Build GRCh38
rs965917635 Chr.9: 4662400-4662400 on Build GRCh38
rs776628224 Chr.9: 4662417-4662417 on Build GRCh38
rs749885838 Chr.9: 4662471-4662471 on Build GRCh38
rs980078358 Chr.9: 4662462-4662462 on Build GRCh38
rs745374841 Chr.9: 4662386-4662386 on Build GRCh38
rs1048763629 Chr.9: 4662373-4662373 on Build GRCh38
rs975025512 Chr.9: 4662424-4662424 on Build GRCh38
rs766410645 Chr.9: 4662364-4662364 on Build GRCh38
rs947662458 Chr.9: 4662341-4662341 on Build GRCh38
rs760841863 Chr.9: 4662410-4662410 on Build GRCh38
rs866381886 Chr.9: 4662510-4662510 on Build GRCh38
rs777546834 Chr.9: 4662344-4662344 on Build GRCh38
rs553329220 Chr.9: 4662423-4662423 on Build GRCh38
rs779821701 Chr.9: 4662479-4662479 on Build GRCh38
rs771158932 Chr.9: 4662416-4662416 on Build GRCh38
rs908034218 Chr.9: 4662470-4662470 on Build GRCh38
rs922160414 Chr.9: 4662343-4662343 on Build GRCh38
rs186498890 Chr.9: 4662537-4662537 on Build GRCh38
rs918836876 Chr.9: 4662481-4662481 on Build GRCh38
rs887536232 Chr.9: 4662377-4662377 on Build GRCh38
rs568620475 Chr.9: 4662357-4662357 on Build GRCh38
rs762236793 Chr.9: 4662478-4662479 on Build GRCh38
rs1021946578 Chr.9: 4662414-4662414 on Build GRCh38
rs777547236 Chr.9: 4662372-4662372 on Build GRCh38
rs1012223972 Chr.9: 4662383-4662383 on Build GRCh38
rs769612565 Chr.9: 4662388-4662388 on Build GRCh38
rs768964167 Chr.9: 4662430-4662432 on Build GRCh38
rs989171683 Chr.9: 4662412-4662412 on Build GRCh38
rs763459517 Chr.9: 4662518-4662518 on Build GRCh38
rs573096039 Chr.9: 4662449-4662449 on Build GRCh38
rs747945489 Chr.9: 4662487-4662487 on Build GRCh38
rs575060963 Chr.9: 4662526-4662526 on Build GRCh38
rs755749721 Chr.9: 4662534-4662534 on Build GRCh38
rs762324944 Chr.9: 4662523-4662523 on Build GRCh38
rs34250374 Chr.9: 4662394-4662394 on Build GRCh38
rs773475238 Chr.9: 4662363-4662363 on Build GRCh38
rs765650862 Chr.9: 4662422-4662422 on Build GRCh38

View Full Product Details