Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 113397156-113397383 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SDS
Entrez Gene ID
10993
Gene Name
serine dehydratase
Gene Aliases
SDH
UniGene
Hs.439023
Gene Chromosome Location
Chr.12: 113392445-113403887 on Build GRCh38
Transcripts
NM_006843.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3416513 c.489C>T p.I163I Chr.12: 113397329-113397329 on Build GRCh38
5667404 c.444C>A p.I148I Chr.12: 113397374-113397374 on Build GRCh38
7120155 c.480G>A p.P160P Chr.12: 113397338-113397338 on Build GRCh38
3456428 c.466C>T p.L156L Chr.12: 113397352-113397352 on Build GRCh38
6994154 c.479C>T p.P160L Chr.12: 113397339-113397339 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs977007363 Chr.12: 113397316-113397316 on Build GRCh38
rs556575776 Chr.12: 113397282-113397282 on Build GRCh38
rs151240589 Chr.12: 113397327-113397327 on Build GRCh38
rs762214578 Chr.12: 113397348-113397348 on Build GRCh38
rs750546750 Chr.12: 113397299-113397299 on Build GRCh38
rs768456672 Chr.12: 113397227-113397227 on Build GRCh38
rs200189238 Chr.12: 113397329-113397329 on Build GRCh38
rs770594760 Chr.12: 113397256-113397256 on Build GRCh38
rs185843116 Chr.12: 113397314-113397314 on Build GRCh38
rs143487976 Chr.12: 113397297-113397297 on Build GRCh38
rs34607566 Chr.12: 113397257-113397257 on Build GRCh38
rs955378757 Chr.12: 113397262-113397262 on Build GRCh38
rs368719444 Chr.12: 113397358-113397358 on Build GRCh38
rs201884808 Chr.12: 113397320-113397320 on Build GRCh38
rs780262006 Chr.12: 113397200-113397200 on Build GRCh38
rs777637583 Chr.12: 113397326-113397326 on Build GRCh38
rs780172339 Chr.12: 113397310-113397310 on Build GRCh38
rs750522334 Chr.12: 113397177-113397177 on Build GRCh38
rs545130942 Chr.12: 113397350-113397350 on Build GRCh38
rs755983426 Chr.12: 113397279-113397279 on Build GRCh38
rs762480559 Chr.12: 113397283-113397283 on Build GRCh38
rs759076665 Chr.12: 113397259-113397259 on Build GRCh38
rs780817430 Chr.12: 113397244-113397244 on Build GRCh38
rs773817124 Chr.12: 113397351-113397351 on Build GRCh38
rs771623291 Chr.12: 113397338-113397338 on Build GRCh38
rs759287194 Chr.12: 113397331-113397331 on Build GRCh38
rs752509823 Chr.12: 113397199-113397200 on Build GRCh38
rs888575832 Chr.12: 113397180-113397180 on Build GRCh38
rs1003988026 Chr.12: 113397194-113397194 on Build GRCh38
rs777061581 Chr.12: 113397339-113397339 on Build GRCh38
rs758695888 Chr.12: 113397199-113397199 on Build GRCh38
rs762393608 Chr.12: 113397251-113397251 on Build GRCh38
rs746801283 Chr.12: 113397211-113397211 on Build GRCh38
rs1010774727 Chr.12: 113397255-113397255 on Build GRCh38
rs371872753 Chr.12: 113397313-113397313 on Build GRCh38
rs779714832 Chr.12: 113397311-113397311 on Build GRCh38
rs771594839 Chr.12: 113397212-113397212 on Build GRCh38
rs772715165 Chr.12: 113397247-113397247 on Build GRCh38
rs764565250 Chr.12: 113397272-113397272 on Build GRCh38
rs769736002 Chr.12: 113397323-113397323 on Build GRCh38
rs770465764 Chr.12: 113397328-113397328 on Build GRCh38
rs985781406 Chr.12: 113397289-113397289 on Build GRCh38
rs758505126 Chr.12: 113397302-113397302 on Build GRCh38
rs763929061 Chr.12: 113397314-113397315 on Build GRCh38
rs146027909 Chr.12: 113397245-113397245 on Build GRCh38
rs766530653 Chr.12: 113397363-113397363 on Build GRCh38

View Full Product Details