Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 123638111-123638384 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCN3B
Entrez Gene ID
55800
Gene Name
sodium voltage-gated channel beta subunit 3
Gene Aliases
ATFB16, BRGDA7, HSA243396, SCNB3
UniGene
Hs.4865
Gene Chromosome Location
Chr.11: 123629187-123654607 on Build GRCh38
Transcripts
NM_001040151.1 NM_018400.3 XM_011542897.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4829307 c.584C>T p.A195V Chr.11: 123638186-123638186 on Build GRCh38
3666147 c.487T>C p.Y163H Chr.11: 123638283-123638283 on Build GRCh38
7333505 c.561C>A p.A187A Chr.11: 123638209-123638209 on Build GRCh38
1352180 c.561C>T p.A187A Chr.11: 123638209-123638209 on Build GRCh38
4805242 c.584C>A p.A195E Chr.11: 123638186-123638186 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148226737 Chr.11: 123638321-123638321 on Build GRCh38
rs753068381 Chr.11: 123638284-123638284 on Build GRCh38
rs34964168 Chr.11: 123638188-123638188 on Build GRCh38
rs7112258 Chr.11: 123638134-123638134 on Build GRCh38
rs763585108 Chr.11: 123638151-123638151 on Build GRCh38
rs752001951 Chr.11: 123638157-123638157 on Build GRCh38
rs768340445 Chr.11: 123638225-123638225 on Build GRCh38
rs1001532028 Chr.11: 123638281-123638281 on Build GRCh38
rs764280230 Chr.11: 123638318-123638318 on Build GRCh38
rs375755770 Chr.11: 123638187-123638187 on Build GRCh38
rs774317879 Chr.11: 123638327-123638330 on Build GRCh38
rs896486186 Chr.11: 123638251-123638251 on Build GRCh38
rs771620122 Chr.11: 123638247-123638247 on Build GRCh38
rs767843514 Chr.11: 123638186-123638186 on Build GRCh38
rs780983107 Chr.11: 123638229-123638229 on Build GRCh38
rs774712002 Chr.11: 123638250-123638250 on Build GRCh38
rs772885993 Chr.11: 123638245-123638245 on Build GRCh38
rs755281336 Chr.11: 123638168-123638168 on Build GRCh38
rs747776064 Chr.11: 123638233-123638233 on Build GRCh38
rs766097713 Chr.11: 123638309-123638315 on Build GRCh38
rs587777557 Chr.11: 123638288-123638288 on Build GRCh38
rs374500940 Chr.11: 123638146-123638146 on Build GRCh38
rs753978888 Chr.11: 123638348-123638348 on Build GRCh38
rs767798639 Chr.11: 123638260-123638260 on Build GRCh38
rs571255893 Chr.11: 123638286-123638286 on Build GRCh38
rs1033907865 Chr.11: 123638287-123638287 on Build GRCh38
rs760034688 Chr.11: 123638253-123638253 on Build GRCh38
rs1037043071 Chr.11: 123638354-123638354 on Build GRCh38
rs939982801 Chr.11: 123638282-123638282 on Build GRCh38
rs547718216 Chr.11: 123638237-123638237 on Build GRCh38
rs769530589 Chr.11: 123638244-123638244 on Build GRCh38
rs766959283 Chr.11: 123638154-123638154 on Build GRCh38
rs769173831 Chr.11: 123638208-123638208 on Build GRCh38
rs757324218 Chr.11: 123638357-123638357 on Build GRCh38
rs575419664 Chr.11: 123638200-123638200 on Build GRCh38
rs746888620 Chr.11: 123638209-123638209 on Build GRCh38
rs527845801 Chr.11: 123638331-123638331 on Build GRCh38

View Full Product Details