Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 140283836-140284073 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRAPPC9
Entrez Gene ID
83696
Gene Name
trafficking protein particle complex 9
Gene Aliases
IBP, IKBKBBP, MRT13, NIBP, T1, TRS120
UniGene
Hs.654911
Gene Chromosome Location
Chr.8: 139727725-140458579 on Build GRCh38
Transcripts
NM_001160372.3 NM_001321646.1 NM_031466.7 XM_011517326.2 XM_011517328.2 XM_011517330.2 XM_017013893.1 XM_017013894.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1674208 c.2404C>T p.P802S Chr.8: 140283893-140283893 on Build GRCh38
4365312 c.2359G>A p.E787K Chr.8: 140283938-140283938 on Build GRCh38
4957491 c.2401C>A p.L801M Chr.8: 140283896-140283896 on Build GRCh38
4961370 c.2371G>A p.A791T Chr.8: 140283926-140283926 on Build GRCh38
6807566 c.2332G>A p.G778R Chr.8: 140283965-140283965 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765243773 Chr.8: 140284025-140284025 on Build GRCh38
rs761179684 Chr.8: 140283927-140283927 on Build GRCh38
rs374990793 Chr.8: 140283879-140283879 on Build GRCh38
rs375784438 Chr.8: 140284008-140284008 on Build GRCh38
rs749228603 Chr.8: 140284051-140284051 on Build GRCh38
rs376768296 Chr.8: 140283906-140283906 on Build GRCh38
rs376735305 Chr.8: 140283967-140283967 on Build GRCh38
rs768331586 Chr.8: 140283918-140283918 on Build GRCh38
rs372142706 Chr.8: 140283896-140283896 on Build GRCh38
rs199623517 Chr.8: 140283939-140283939 on Build GRCh38
rs748405461 Chr.8: 140283890-140283890 on Build GRCh38
rs754983621 Chr.8: 140284054-140284054 on Build GRCh38
rs74414386 Chr.8: 140284041-140284041 on Build GRCh38
rs371880751 Chr.8: 140283966-140283966 on Build GRCh38
rs755771223 Chr.8: 140284040-140284040 on Build GRCh38
rs147687394 Chr.8: 140284001-140284001 on Build GRCh38
rs760093468 Chr.8: 140284021-140284021 on Build GRCh38
rs949962880 Chr.8: 140283936-140283936 on Build GRCh38
rs1017127961 Chr.8: 140283923-140283923 on Build GRCh38
rs773574715 Chr.8: 140284006-140284006 on Build GRCh38
rs201556589 Chr.8: 140284009-140284009 on Build GRCh38
rs368512631 Chr.8: 140283877-140283877 on Build GRCh38
rs370044850 Chr.8: 140283977-140283977 on Build GRCh38
rs200235924 Chr.8: 140283924-140283924 on Build GRCh38
rs749881435 Chr.8: 140283936-140283936 on Build GRCh38
rs372777496 Chr.8: 140283878-140283878 on Build GRCh38
rs145138747 Chr.8: 140283898-140283898 on Build GRCh38
rs762215508 Chr.8: 140283857-140283857 on Build GRCh38
rs141531887 Chr.8: 140283912-140283912 on Build GRCh38
rs147044768 Chr.8: 140283926-140283926 on Build GRCh38
rs201157726 Chr.8: 140283858-140283858 on Build GRCh38
rs902108618 Chr.8: 140283873-140283873 on Build GRCh38
rs778422621 Chr.8: 140284055-140284055 on Build GRCh38
rs780062048 Chr.8: 140284050-140284050 on Build GRCh38
rs535577825 Chr.8: 140283978-140283978 on Build GRCh38
rs946098801 Chr.8: 140284002-140284002 on Build GRCh38
rs1025984933 Chr.8: 140284021-140284021 on Build GRCh38
rs369629465 Chr.8: 140283917-140283917 on Build GRCh38
rs754388457 Chr.8: 140283938-140283938 on Build GRCh38
rs780972944 Chr.8: 140283958-140283958 on Build GRCh38
rs756615054 Chr.8: 140283921-140283921 on Build GRCh38
rs752950082 Chr.8: 140283947-140283947 on Build GRCh38
rs758562541 Chr.8: 140283955-140283955 on Build GRCh38
rs780906963 Chr.8: 140283900-140283900 on Build GRCh38
rs950450278 Chr.8: 140284034-140284034 on Build GRCh38
rs765364454 Chr.8: 140283946-140283946 on Build GRCh38
rs373886400 Chr.8: 140283993-140283993 on Build GRCh38
rs750939917 Chr.8: 140283868-140283868 on Build GRCh38
rs756031831 Chr.8: 140283969-140283969 on Build GRCh38
rs764246926 Chr.8: 140284031-140284031 on Build GRCh38
rs751858973 Chr.8: 140284037-140284037 on Build GRCh38
rs191363774 Chr.8: 140283865-140283865 on Build GRCh38
rs767914895 Chr.8: 140283925-140283925 on Build GRCh38
rs867224801 Chr.8: 140283973-140283973 on Build GRCh38
rs768280568 Chr.8: 140283992-140283992 on Build GRCh38
rs779081698 Chr.8: 140283880-140283880 on Build GRCh38
rs758604818 Chr.8: 140284030-140284030 on Build GRCh38
rs752722432 Chr.8: 140284028-140284028 on Build GRCh38
rs766891466 Chr.8: 140283931-140283931 on Build GRCh38
rs778380217 Chr.8: 140283972-140283972 on Build GRCh38
rs200557642 Chr.8: 140284015-140284015 on Build GRCh38

View Full Product Details