Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 7039586-7039857 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF144A
Entrez Gene ID
9781
Gene Name
ring finger protein 144A
Gene Aliases
RNF144, UBCE7IP4
UniGene
Hs.22146
Gene Chromosome Location
Chr.2: 6917392-7076886 on Build GRCh38
Transcripts
NM_014746.4 XM_005246200.3 XM_005246202.4 XM_017005396.1 XM_017005397.1 XM_017005398.1 XM_017005399.1 XM_017005400.1 XM_017005401.1 XM_017005402.1 XM_017005403.1 XM_017005404.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5344253 c.858C>T p.D286D Chr.2: 7039739-7039739 on Build GRCh38
3737628 c.878A>G p.*293W Chr.2: 7039759-7039759 on Build GRCh38
4095496 c.872C>A p.P291H Chr.2: 7039753-7039753 on Build GRCh38
3729880 c.757_758insT p.A255fs*30 Chr.2: 7039638-7039639 on Build GRCh38
4095495 c.755G>A p.G252D Chr.2: 7039636-7039636 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144778745 Chr.2: 7039724-7039724 on Build GRCh38
rs772351212 Chr.2: 7039809-7039809 on Build GRCh38
rs748863431 Chr.2: 7039778-7039778 on Build GRCh38
rs368482595 Chr.2: 7039767-7039767 on Build GRCh38
rs776205017 Chr.2: 7039671-7039671 on Build GRCh38
rs760053213 Chr.2: 7039795-7039795 on Build GRCh38
rs747788326 Chr.2: 7039770-7039770 on Build GRCh38
rs192378723 Chr.2: 7039827-7039827 on Build GRCh38
rs753914440 Chr.2: 7039742-7039742 on Build GRCh38
rs1041463523 Chr.2: 7039768-7039768 on Build GRCh38
rs1026107236 Chr.2: 7039673-7039673 on Build GRCh38
rs752947866 Chr.2: 7039618-7039618 on Build GRCh38
rs770412959 Chr.2: 7039780-7039780 on Build GRCh38
rs746719592 Chr.2: 7039658-7039658 on Build GRCh38
rs1050065448 Chr.2: 7039815-7039815 on Build GRCh38
rs921152907 Chr.2: 7039821-7039821 on Build GRCh38
rs758365664 Chr.2: 7039638-7039638 on Build GRCh38
rs1014492820 Chr.2: 7039617-7039617 on Build GRCh38
rs766805474 Chr.2: 7039711-7039711 on Build GRCh38
rs201740884 Chr.2: 7039794-7039794 on Build GRCh38
rs758418992 Chr.2: 7039743-7039743 on Build GRCh38
rs769335180 Chr.2: 7039694-7039694 on Build GRCh38
rs568338318 Chr.2: 7039769-7039769 on Build GRCh38
rs72783732 Chr.2: 7039830-7039830 on Build GRCh38
rs971365658 Chr.2: 7039691-7039691 on Build GRCh38
rs373649202 Chr.2: 7039771-7039771 on Build GRCh38
rs745835992 Chr.2: 7039637-7039637 on Build GRCh38
rs374871027 Chr.2: 7039727-7039727 on Build GRCh38
rs767698220 Chr.2: 7039614-7039614 on Build GRCh38
rs756316410 Chr.2: 7039739-7039739 on Build GRCh38
rs780010880 Chr.2: 7039648-7039648 on Build GRCh38
rs768495307 Chr.2: 7039668-7039668 on Build GRCh38
rs927193239 Chr.2: 7039719-7039719 on Build GRCh38
rs747815989 Chr.2: 7039688-7039688 on Build GRCh38
rs777268135 Chr.2: 7039775-7039775 on Build GRCh38
rs756194391 Chr.2: 7039625-7039625 on Build GRCh38
rs772658335 Chr.2: 7039697-7039697 on Build GRCh38
rs759872373 Chr.2: 7039725-7039725 on Build GRCh38
rs751502609 Chr.2: 7039754-7039754 on Build GRCh38
rs534776470 Chr.2: 7039700-7039700 on Build GRCh38
rs116762840 Chr.2: 7039748-7039748 on Build GRCh38
rs937233696 Chr.2: 7039726-7039726 on Build GRCh38
rs1007114869 Chr.2: 7039828-7039828 on Build GRCh38
rs752900255 Chr.2: 7039734-7039734 on Build GRCh38

View Full Product Details