Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 114938065-114938336 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRUB1
Entrez Gene ID
142940
Gene Name
TruB pseudouridine synthase family member 1
Gene Aliases
PUS4
UniGene
Hs.21187
Gene Chromosome Location
Chr.10: 114938193-114977680 on Build GRCh38
Transcripts
NM_139169.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6229663 c.59C>T p.P20L Chr.10: 114938312-114938312 on Build GRCh38
4011860 c.82G>A p.V28I Chr.10: 114938335-114938335 on Build GRCh38
3434413 c.56C>T p.S19F Chr.10: 114938309-114938309 on Build GRCh38
4957315 c.24G>A p.V8V Chr.10: 114938277-114938277 on Build GRCh38
6065077 c.24G>T p.V8V Chr.10: 114938277-114938277 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777873032 Chr.10: 114938260-114938260 on Build GRCh38
rs369362278 Chr.10: 114938244-114938244 on Build GRCh38
rs192075462 Chr.10: 114938155-114938155 on Build GRCh38
rs576664891 Chr.10: 114938245-114938245 on Build GRCh38
rs748901670 Chr.10: 114938277-114938277 on Build GRCh38
rs777507141 Chr.10: 114938261-114938261 on Build GRCh38
rs750579371 Chr.10: 114938248-114938248 on Build GRCh38
rs781079988 Chr.10: 114938273-114938273 on Build GRCh38
rs758134796 Chr.10: 114938259-114938259 on Build GRCh38
rs200809062 Chr.10: 114938211-114938211 on Build GRCh38
rs1004082062 Chr.10: 114938144-114938144 on Build GRCh38
rs780298786 Chr.10: 114938203-114938203 on Build GRCh38
rs762368141 Chr.10: 114938246-114938246 on Build GRCh38
rs777701287 Chr.10: 114938219-114938219 on Build GRCh38
rs909082336 Chr.10: 114938088-114938088 on Build GRCh38
rs375548422 Chr.10: 114938284-114938284 on Build GRCh38
rs962797701 Chr.10: 114938197-114938197 on Build GRCh38
rs551463217 Chr.10: 114938228-114938228 on Build GRCh38
rs762245615 Chr.10: 114938296-114938296 on Build GRCh38
rs768178093 Chr.10: 114938309-114938309 on Build GRCh38
rs376743642 Chr.10: 114938223-114938223 on Build GRCh38
rs141193231 Chr.10: 114938100-114938100 on Build GRCh38
rs558311749 Chr.10: 114938235-114938235 on Build GRCh38
rs765097169 Chr.10: 114938120-114938120 on Build GRCh38
rs377402974 Chr.10: 114938224-114938224 on Build GRCh38
rs368115884 Chr.10: 114938274-114938274 on Build GRCh38
rs747962323 Chr.10: 114938210-114938210 on Build GRCh38
rs991747728 Chr.10: 114938226-114938226 on Build GRCh38
rs1029604109 Chr.10: 114938112-114938112 on Build GRCh38
rs751762173 Chr.10: 114938311-114938311 on Build GRCh38
rs545293079 Chr.10: 114938184-114938184 on Build GRCh38
rs1015434054 Chr.10: 114938174-114938174 on Build GRCh38
rs1054888014 Chr.10: 114938096-114938096 on Build GRCh38
rs756215966 Chr.10: 114938249-114938249 on Build GRCh38
rs765682149 Chr.10: 114938213-114938216 on Build GRCh38
rs769375045 Chr.10: 114938227-114938227 on Build GRCh38
rs1045702801 Chr.10: 114938182-114938182 on Build GRCh38
rs754035471 Chr.10: 114938258-114938258 on Build GRCh38
rs775201648 Chr.10: 114938230-114938230 on Build GRCh38
rs146924418 Chr.10: 114938254-114938254 on Build GRCh38
rs183399437 Chr.10: 114938247-114938247 on Build GRCh38
rs138212899 Chr.10: 114938270-114938270 on Build GRCh38
rs752785055 Chr.10: 114938312-114938313 on Build GRCh38
rs559435727 Chr.10: 114938276-114938276 on Build GRCh38
rs996822000 Chr.10: 114938103-114938103 on Build GRCh38
rs377229377 Chr.10: 114938268-114938268 on Build GRCh38
rs752770081 Chr.10: 114938170-114938170 on Build GRCh38
rs17092618 Chr.10: 114938201-114938201 on Build GRCh38
rs770463703 Chr.10: 114938221-114938221 on Build GRCh38
rs533529707 Chr.10: 114938217-114938217 on Build GRCh38
rs774806190 Chr.10: 114938285-114938285 on Build GRCh38
rs372255913 Chr.10: 114938129-114938129 on Build GRCh38

View Full Product Details