Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 50516410-50516657 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCAPH2
Entrez Gene ID
29781
Gene Name
non-SMC condensin II complex subunit H2
Gene Aliases
CAPH2
UniGene
Hs.730607
Gene Chromosome Location
Chr.22: 50508216-50524780 on Build GRCh38
Transcripts
NM_001185011.1 NM_014551.4 NM_152299.3 XM_005261912.4 XM_011530685.2 XM_017028793.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1216802 c.136G>A p.E46K Chr.22: 50516474-50516474 on Build GRCh38
7269165 c.195C>T p.C65C Chr.22: 50516533-50516533 on Build GRCh38
7269164 c.195C>T p.C65C Chr.22: 50516533-50516533 on Build GRCh38
1035490 c.153G>T p.M51I Chr.22: 50516491-50516491 on Build GRCh38
1593135 c.153G>T p.M51I Chr.22: 50516491-50516491 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781620450 Chr.22: 50516572-50516572 on Build GRCh38
rs769120514 Chr.22: 50516456-50516456 on Build GRCh38
rs28744512 Chr.22: 50516473-50516473 on Build GRCh38
rs770066460 Chr.22: 50516482-50516482 on Build GRCh38
rs764973773 Chr.22: 50516539-50516539 on Build GRCh38
rs570609283 Chr.22: 50516512-50516512 on Build GRCh38
rs770862985 Chr.22: 50516498-50516498 on Build GRCh38
rs764117167 Chr.22: 50516526-50516526 on Build GRCh38
rs758047691 Chr.22: 50516564-50516564 on Build GRCh38
rs572597393 Chr.22: 50516565-50516565 on Build GRCh38
rs568414719 Chr.22: 50516615-50516615 on Build GRCh38
rs904814126 Chr.22: 50516556-50516556 on Build GRCh38
rs748547937 Chr.22: 50516586-50516586 on Build GRCh38
rs747565217 Chr.22: 50516443-50516443 on Build GRCh38
rs755535814 Chr.22: 50516569-50516569 on Build GRCh38
rs539900713 Chr.22: 50516633-50516633 on Build GRCh38
rs753701599 Chr.22: 50516533-50516533 on Build GRCh38
rs763120856 Chr.22: 50516497-50516497 on Build GRCh38
rs748497607 Chr.22: 50516474-50516474 on Build GRCh38
rs147983222 Chr.22: 50516561-50516562 on Build GRCh38
rs1013099927 Chr.22: 50516509-50516509 on Build GRCh38
rs79758616 Chr.22: 50516638-50516638 on Build GRCh38
rs11912147 Chr.22: 50516603-50516603 on Build GRCh38
rs866121965 Chr.22: 50516466-50516466 on Build GRCh38
rs201616514 Chr.22: 50516439-50516439 on Build GRCh38
rs780633992 Chr.22: 50516440-50516440 on Build GRCh38
rs762292159 Chr.22: 50516555-50516555 on Build GRCh38
rs150971534 Chr.22: 50516499-50516499 on Build GRCh38
rs770012065 Chr.22: 50516587-50516587 on Build GRCh38
rs749994483 Chr.22: 50516553-50516553 on Build GRCh38
rs371992143 Chr.22: 50516437-50516437 on Build GRCh38
rs369628738 Chr.22: 50516590-50516590 on Build GRCh38
rs778075051 Chr.22: 50516597-50516597 on Build GRCh38
rs761449979 Chr.22: 50516534-50516534 on Build GRCh38
rs773238653 Chr.22: 50516488-50516488 on Build GRCh38

View Full Product Details