Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 37840027-37840292 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PGM2
Entrez Gene ID
55276
Gene Name
phosphoglucomutase 2
Gene Aliases
MSTP006
UniGene
Hs.23363
Gene Chromosome Location
Chr.4: 37826660-37862938 on Build GRCh38
Transcripts
NM_018290.3 XM_011513711.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7165289 c.640A>G p.S214G Chr.4: 37840180-37840180 on Build GRCh38
733546 c.625G>A p.D209N Chr.4: 37840165-37840165 on Build GRCh38
7090305 c.571G>T p.G191W Chr.4: 37840111-37840111 on Build GRCh38
7227104 c.652C>T p.L218F Chr.4: 37840192-37840192 on Build GRCh38
1055075 c.594A>C p.E198D Chr.4: 37840134-37840134 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs535172295 Chr.4: 37840254-37840254 on Build GRCh38
rs924869783 Chr.4: 37840087-37840087 on Build GRCh38
rs778250303 Chr.4: 37840066-37840066 on Build GRCh38
rs753775155 Chr.4: 37840055-37840055 on Build GRCh38
rs757963215 Chr.4: 37840071-37840071 on Build GRCh38
rs774773425 Chr.4: 37840123-37840123 on Build GRCh38
rs188074789 Chr.4: 37840105-37840105 on Build GRCh38
rs768941882 Chr.4: 37840092-37840092 on Build GRCh38
rs777274812 Chr.4: 37840169-37840169 on Build GRCh38
rs766213593 Chr.4: 37840139-37840139 on Build GRCh38
rs150576315 Chr.4: 37840145-37840145 on Build GRCh38
rs1051320373 Chr.4: 37840177-37840177 on Build GRCh38
rs770172003 Chr.4: 37840202-37840202 on Build GRCh38
rs574168507 Chr.4: 37840084-37840084 on Build GRCh38
rs201966949 Chr.4: 37840258-37840258 on Build GRCh38
rs761578571 Chr.4: 37840178-37840179 on Build GRCh38
rs746606591 Chr.4: 37840179-37840179 on Build GRCh38
rs767877668 Chr.4: 37840264-37840264 on Build GRCh38
rs148083154 Chr.4: 37840093-37840093 on Build GRCh38
rs547953369 Chr.4: 37840192-37840192 on Build GRCh38
rs757988566 Chr.4: 37840165-37840165 on Build GRCh38
rs761008942 Chr.4: 37840088-37840088 on Build GRCh38
rs762089923 Chr.4: 37840257-37840257 on Build GRCh38
rs749315628 Chr.4: 37840217-37840217 on Build GRCh38
rs566471580 Chr.4: 37840197-37840197 on Build GRCh38
rs977162803 Chr.4: 37840108-37840108 on Build GRCh38
rs776570516 Chr.4: 37840114-37840114 on Build GRCh38
rs934269477 Chr.4: 37840170-37840170 on Build GRCh38
rs768726659 Chr.4: 37840219-37840219 on Build GRCh38
rs747366554 Chr.4: 37840089-37840089 on Build GRCh38
rs940153653 Chr.4: 37840214-37840214 on Build GRCh38
rs565494422 Chr.4: 37840106-37840106 on Build GRCh38
rs777533193 Chr.4: 37840250-37840250 on Build GRCh38
rs929292636 Chr.4: 37840190-37840190 on Build GRCh38
rs776132216 Chr.4: 37840206-37840206 on Build GRCh38
rs756905605 Chr.4: 37840181-37840181 on Build GRCh38
rs370897461 Chr.4: 37840119-37840119 on Build GRCh38
rs867780120 Chr.4: 37840240-37840240 on Build GRCh38
rs144125166 Chr.4: 37840104-37840104 on Build GRCh38
rs199867267 Chr.4: 37840057-37840057 on Build GRCh38
rs955080930 Chr.4: 37840062-37840062 on Build GRCh38
rs776992624 Chr.4: 37840099-37840099 on Build GRCh38
rs369810311 Chr.4: 37840127-37840127 on Build GRCh38
rs113775951 Chr.4: 37840094-37840094 on Build GRCh38
rs747713507 Chr.4: 37840070-37840070 on Build GRCh38
rs774581539 Chr.4: 37840221-37840221 on Build GRCh38
rs750611272 Chr.4: 37840164-37840164 on Build GRCh38
rs138680388 Chr.4: 37840151-37840151 on Build GRCh38
rs922324369 Chr.4: 37840059-37840059 on Build GRCh38
rs562869938 Chr.4: 37840101-37840101 on Build GRCh38

View Full Product Details