Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 53097679-53097910 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGFBP6
Entrez Gene ID
3489
Gene Name
insulin like growth factor binding protein 6
Gene Aliases
IBP6
UniGene
Hs.274313
Gene Chromosome Location
Chr.12: 53097503-53102344 on Build GRCh38
Transcripts
NM_002178.2 XM_017019264.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6275895 c.27G>T p.P9P Chr.12: 53097744-53097744 on Build GRCh38
5505345 c.30G>A p.L10L Chr.12: 53097747-53097747 on Build GRCh38
6367961 c.133G>T p.V45L Chr.12: 53097850-53097850 on Build GRCh38
5555329 c.99G>A p.G33G Chr.12: 53097816-53097816 on Build GRCh38
6416614 c.55G>A p.A19T Chr.12: 53097772-53097772 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746720408 Chr.12: 53097707-53097707 on Build GRCh38
rs559725416 Chr.12: 53097809-53097809 on Build GRCh38
rs913609172 Chr.12: 53097842-53097842 on Build GRCh38
rs112479506 Chr.12: 53097727-53097727 on Build GRCh38
rs940728756 Chr.12: 53097832-53097832 on Build GRCh38
rs776237463 Chr.12: 53097836-53097836 on Build GRCh38
rs768996992 Chr.12: 53097739-53097739 on Build GRCh38
rs760925481 Chr.12: 53097803-53097803 on Build GRCh38
rs17123156 Chr.12: 53097747-53097747 on Build GRCh38
rs759385626 Chr.12: 53097866-53097866 on Build GRCh38
rs769965272 Chr.12: 53097837-53097837 on Build GRCh38
rs765013483 Chr.12: 53097870-53097870 on Build GRCh38
rs748160271 Chr.12: 53097756-53097756 on Build GRCh38
rs925102041 Chr.12: 53097757-53097757 on Build GRCh38
rs55922809 Chr.12: 53097876-53097876 on Build GRCh38
rs749537120 Chr.12: 53097744-53097746 on Build GRCh38
rs773401386 Chr.12: 53097798-53097798 on Build GRCh38
rs969252229 Chr.12: 53097746-53097746 on Build GRCh38
rs757972739 Chr.12: 53097882-53097882 on Build GRCh38
rs796302652 Chr.12: 53097805-53097805 on Build GRCh38
rs547855662 Chr.12: 53097784-53097784 on Build GRCh38
rs1045127931 Chr.12: 53097881-53097881 on Build GRCh38
rs777567670 Chr.12: 53097699-53097699 on Build GRCh38
rs772197160 Chr.12: 53097771-53097771 on Build GRCh38
rs1014635779 Chr.12: 53097706-53097706 on Build GRCh38
rs770929787 Chr.12: 53097743-53097744 on Build GRCh38
rs914364078 Chr.12: 53097700-53097700 on Build GRCh38

View Full Product Details