Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 44119292-44119537 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC102724159
Entrez Gene ID
102724159
Gene Name
periodic tryptophan protein 2 homolog
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.21: 5130871-5154767 on Build GRCh38
Transcripts
XM_006723910.1 NM_005049.2 XM_011529667.2

Gene Symbol
PWP2
Entrez Gene ID
5822
Gene Name
PWP2 periodic tryptophan protein homolog (yeast)
Gene Aliases
EHOC-17, PWP2H, UTP1
UniGene
Hs.449076
Gene Chromosome Location
Chr.21: 44107262-44131181 on Build GRCh38
Transcripts
XM_006723910.1 NM_005049.2 XM_011529667.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4843440 c.1153G>A p.G385R Chr.21: 44119488-44119488 on Build GRCh38
5568218 c.1146G>A p.S382S Chr.21: 44119481-44119481 on Build GRCh38
4593590 c.1178A>G p.D393G Chr.21: 44119513-44119513 on Build GRCh38
5712966 c.1179C>T p.D393D Chr.21: 44119514-44119514 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs765144760 Chr.21: 44119484-44119484 on Build GRCh38
rs571856443 Chr.21: 44119349-44119349 on Build GRCh38
rs770012249 Chr.21: 44119350-44119350 on Build GRCh38
rs143110108 Chr.21: 44119481-44119481 on Build GRCh38
rs138472662 Chr.21: 44119480-44119480 on Build GRCh38
rs535568001 Chr.21: 44119517-44119517 on Build GRCh38
rs752681930 Chr.21: 44119366-44119366 on Build GRCh38
rs781458113 Chr.21: 44119346-44119346 on Build GRCh38
rs148758953 Chr.21: 44119499-44119499 on Build GRCh38
rs529411997 Chr.21: 44119344-44119344 on Build GRCh38
rs775248369 Chr.21: 44119487-44119487 on Build GRCh38
rs557366551 Chr.21: 44119469-44119469 on Build GRCh38
rs764894517 Chr.21: 44119365-44119365 on Build GRCh38
rs930017728 Chr.21: 44119384-44119384 on Build GRCh38
rs145949707 Chr.21: 44119430-44119430 on Build GRCh38
rs375459760 Chr.21: 44119415-44119415 on Build GRCh38
rs2855652 Chr.21: 44119359-44119359 on Build GRCh38
rs536959317 Chr.21: 44119482-44119482 on Build GRCh38
rs374556467 Chr.21: 44119358-44119358 on Build GRCh38
rs751459136 Chr.21: 44119513-44119513 on Build GRCh38
rs149192996 Chr.21: 44119339-44119339 on Build GRCh38
rs747294969 Chr.21: 44119343-44119343 on Build GRCh38
rs201067049 Chr.21: 44119431-44119431 on Build GRCh38
rs781508053 Chr.21: 44119404-44119404 on Build GRCh38
rs763497984 Chr.21: 44119356-44119356 on Build GRCh38
rs756512750 Chr.21: 44119427-44119427 on Build GRCh38
rs543810839 Chr.21: 44119314-44119314 on Build GRCh38
rs757510017 Chr.21: 44119397-44119397 on Build GRCh38
rs552546557 Chr.21: 44119379-44119379 on Build GRCh38
rs758324282 Chr.21: 44119368-44119368 on Build GRCh38
rs753484980 Chr.21: 44119333-44119333 on Build GRCh38
rs1037572120 Chr.21: 44119461-44119461 on Build GRCh38
rs868200504 Chr.21: 44119326-44119326 on Build GRCh38
rs117894185 Chr.21: 44119360-44119360 on Build GRCh38
rs776085338 Chr.21: 44119355-44119355 on Build GRCh38
rs757349457 Chr.21: 44119514-44119514 on Build GRCh38
rs777760423 Chr.21: 44119342-44119342 on Build GRCh38
rs1019979415 Chr.21: 44119511-44119511 on Build GRCh38
rs777972184 Chr.21: 44119377-44119377 on Build GRCh38
rs574646908 Chr.21: 44119438-44119438 on Build GRCh38
rs749627278 Chr.21: 44119434-44119434 on Build GRCh38
rs746491106 Chr.21: 44119451-44119451 on Build GRCh38
rs746026714 Chr.21: 44119408-44119408 on Build GRCh38
rs954421953 Chr.21: 44119320-44119320 on Build GRCh38
rs371288987 Chr.21: 44119357-44119357 on Build GRCh38
rs868045593 Chr.21: 44119387-44119387 on Build GRCh38
rs750664184 Chr.21: 44119518-44119518 on Build GRCh38
rs144390937 Chr.21: 44119497-44119497 on Build GRCh38

View Full Product Details