Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44771553-44771818 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM11
Entrez Gene ID
4185
Gene Name
ADAM metallopeptidase domain 11
Gene Aliases
MDC
UniGene
Hs.6088
Gene Chromosome Location
Chr.17: 44759037-44781846 on Build GRCh38
Transcripts
NM_001318933.1 NM_002390.5 XM_005257373.3 XM_017024647.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4186452 c.472G>A p.V158I Chr.17: 44771760-44771760 on Build GRCh38
158881 c.419G>A p.R140Q Chr.17: 44771621-44771621 on Build GRCh38
4659259 c.390A>C p.G130G Chr.17: 44771592-44771592 on Build GRCh38
7163605 c.447C>G p.L149L Chr.17: 44771649-44771649 on Build GRCh38
6390467 c.402C>A p.Y134* Chr.17: 44771604-44771604 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766830165 Chr.17: 44771777-44771777 on Build GRCh38
rs540374228 Chr.17: 44771653-44771653 on Build GRCh38
rs142131788 Chr.17: 44771593-44771593 on Build GRCh38
rs753917230 Chr.17: 44771781-44771781 on Build GRCh38
rs747906784 Chr.17: 44771716-44771716 on Build GRCh38
rs374275292 Chr.17: 44771712-44771712 on Build GRCh38
rs760315607 Chr.17: 44771641-44771641 on Build GRCh38
rs771905587 Chr.17: 44771634-44771634 on Build GRCh38
rs548503019 Chr.17: 44771682-44771682 on Build GRCh38
rs929084082 Chr.17: 44771686-44771686 on Build GRCh38
rs751906080 Chr.17: 44771574-44771574 on Build GRCh38
rs750977685 Chr.17: 44771774-44771774 on Build GRCh38
rs765978494 Chr.17: 44771643-44771643 on Build GRCh38
rs773223622 Chr.17: 44771640-44771640 on Build GRCh38
rs755185402 Chr.17: 44771714-44771714 on Build GRCh38
rs778861401 Chr.17: 44771661-44771661 on Build GRCh38
rs750925447 Chr.17: 44771701-44771701 on Build GRCh38
rs779121981 Chr.17: 44771715-44771715 on Build GRCh38
rs757738487 Chr.17: 44771575-44771575 on Build GRCh38
rs746908261 Chr.17: 44771727-44771727 on Build GRCh38
rs147818264 Chr.17: 44771790-44771790 on Build GRCh38
rs776315863 Chr.17: 44771728-44771728 on Build GRCh38
rs769852589 Chr.17: 44771587-44771587 on Build GRCh38
rs774037102 Chr.17: 44771630-44771630 on Build GRCh38
rs114458198 Chr.17: 44771726-44771726 on Build GRCh38
rs898905561 Chr.17: 44771678-44771678 on Build GRCh38
rs771883578 Chr.17: 44771719-44771719 on Build GRCh38
rs371615985 Chr.17: 44771708-44771708 on Build GRCh38
rs755025015 Chr.17: 44771782-44771782 on Build GRCh38
rs762152759 Chr.17: 44771748-44771748 on Build GRCh38
rs1013895185 Chr.17: 44771780-44771780 on Build GRCh38
rs547929486 Chr.17: 44771754-44771754 on Build GRCh38
rs200440815 Chr.17: 44771616-44771616 on Build GRCh38
rs767836498 Chr.17: 44771759-44771759 on Build GRCh38
rs749481116 Chr.17: 44771621-44771621 on Build GRCh38
rs373446189 Chr.17: 44771631-44771631 on Build GRCh38
rs781676844 Chr.17: 44771576-44771576 on Build GRCh38
rs768544543 Chr.17: 44771627-44771627 on Build GRCh38
rs749316218 Chr.17: 44771713-44771713 on Build GRCh38
rs745962413 Chr.17: 44771577-44771577 on Build GRCh38
rs775195920 Chr.17: 44771739-44771739 on Build GRCh38
rs202093736 Chr.17: 44771688-44771688 on Build GRCh38
rs968066115 Chr.17: 44771725-44771725 on Build GRCh38
rs764812732 Chr.17: 44771732-44771732 on Build GRCh38
rs779058125 Chr.17: 44771789-44771789 on Build GRCh38
rs776476231 Chr.17: 44771644-44771644 on Build GRCh38
rs1025609686 Chr.17: 44771787-44771787 on Build GRCh38
rs756636001 Chr.17: 44771775-44771775 on Build GRCh38
rs767968577 Chr.17: 44771690-44771690 on Build GRCh38

View Full Product Details