Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 84176417-84176644 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf99
Entrez Gene ID
387695
Gene Name
chromosome 10 open reading frame 99
Gene Aliases
AP-57, CSBF, UNQ1833
UniGene
Hs.298713
Gene Chromosome Location
Chr.10: 84173764-84185294 on Build GRCh38
Transcripts
NM_207373.2 XM_005269811.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1603791 c.80G>A p.R27H Chr.10: 84176489-84176489 on Build GRCh38
7357962 c.101G>T p.G34V Chr.10: 84176510-84176510 on Build GRCh38
5029749 c.89A>G p.K30R Chr.10: 84176498-84176498 on Build GRCh38
920875 c.79C>T p.R27C Chr.10: 84176488-84176488 on Build GRCh38
5371208 c.90G>A p.K30K Chr.10: 84176499-84176499 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201084865 Chr.10: 84176507-84176507 on Build GRCh38
rs751782604 Chr.10: 84176620-84176620 on Build GRCh38
rs755997018 Chr.10: 84176449-84176449 on Build GRCh38
rs150504515 Chr.10: 84176489-84176489 on Build GRCh38
rs755096491 Chr.10: 84176553-84176553 on Build GRCh38
rs200924237 Chr.10: 84176516-84176516 on Build GRCh38
rs1028941869 Chr.10: 84176606-84176606 on Build GRCh38
rs776829410 Chr.10: 84176501-84176501 on Build GRCh38
rs747127593 Chr.10: 84176480-84176480 on Build GRCh38
rs771418002 Chr.10: 84176581-84176581 on Build GRCh38
rs767530379 Chr.10: 84176440-84176440 on Build GRCh38
rs775162618 Chr.10: 84176537-84176537 on Build GRCh38
rs140646223 Chr.10: 84176547-84176547 on Build GRCh38
rs769157441 Chr.10: 84176506-84176506 on Build GRCh38
rs781064514 Chr.10: 84176555-84176555 on Build GRCh38
rs745985568 Chr.10: 84176577-84176577 on Build GRCh38
rs753682017 Chr.10: 84176544-84176544 on Build GRCh38
rs764058896 Chr.10: 84176601-84176601 on Build GRCh38
rs777362890 Chr.10: 84176450-84176450 on Build GRCh38
rs775634302 Chr.10: 84176609-84176610 on Build GRCh38
rs770201414 Chr.10: 84176492-84176492 on Build GRCh38
rs73315862 Chr.10: 84176586-84176586 on Build GRCh38
rs771262901 Chr.10: 84176488-84176488 on Build GRCh38
rs905256190 Chr.10: 84176582-84176582 on Build GRCh38
rs370200637 Chr.10: 84176534-84176534 on Build GRCh38
rs771642084 Chr.10: 84176587-84176587 on Build GRCh38
rs770136382 Chr.10: 84176603-84176608 on Build GRCh38
rs773675793 Chr.10: 84176505-84176505 on Build GRCh38
rs770373058 Chr.10: 84176592-84176592 on Build GRCh38
rs1003717182 Chr.10: 84176584-84176584 on Build GRCh38
rs780233448 Chr.10: 84176479-84176479 on Build GRCh38
rs927650082 Chr.10: 84176495-84176495 on Build GRCh38
rs376501525 Chr.10: 84176481-84176481 on Build GRCh38
rs996243234 Chr.10: 84176603-84176603 on Build GRCh38
rs192367195 Chr.10: 84176618-84176618 on Build GRCh38
rs750673880 Chr.10: 84176451-84176451 on Build GRCh38
rs553963691 Chr.10: 84176539-84176539 on Build GRCh38
rs776541984 Chr.10: 84176609-84176609 on Build GRCh38
rs185089501 Chr.10: 84176464-84176464 on Build GRCh38
rs954747526 Chr.10: 84176608-84176608 on Build GRCh38
rs756533312 Chr.10: 84176557-84176557 on Build GRCh38
rs367750271 Chr.10: 84176550-84176550 on Build GRCh38
rs374495251 Chr.10: 84176536-84176536 on Build GRCh38
rs144481604 Chr.10: 84176549-84176549 on Build GRCh38
rs147830237 Chr.10: 84176552-84176552 on Build GRCh38
rs752573496 Chr.10: 84176441-84176441 on Build GRCh38
rs141285554 Chr.10: 84176556-84176556 on Build GRCh38
rs774116875 Chr.10: 84176522-84176522 on Build GRCh38
rs533648292 Chr.10: 84176558-84176558 on Build GRCh38
rs749714713 Chr.10: 84176565-84176565 on Build GRCh38
rs761651037 Chr.10: 84176616-84176616 on Build GRCh38
rs1045187659 Chr.10: 84176577-84176577 on Build GRCh38
rs375312628 Chr.10: 84176594-84176594 on Build GRCh38

View Full Product Details