Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 15269688-15269945 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB9
Entrez Gene ID
140462
Gene Name
ankyrin repeat and SOCS box containing 9
Gene Aliases
-
UniGene
Hs.19404
Gene Chromosome Location
Chr.X: 15243987-15270476 on Build GRCh38
Transcripts
NM_001031739.2 NM_001168530.1 NM_001168531.1 NM_024087.2 XM_005274446.1 XM_011545458.1 XM_011545460.1 XM_017029283.1 XM_017029284.1 XM_017029285.1 XM_017029286.1

SNP Detail

SNP ID Chromosome Location
rs959719222 Chr.X: 15269748-15269748 on Build GRCh38
rs937831998 Chr.X: 15269718-15269718 on Build GRCh38
rs140406684 Chr.X: 15269791-15269791 on Build GRCh38
rs769943633 Chr.X: 15269802-15269802 on Build GRCh38
rs150401329 Chr.X: 15269824-15269824 on Build GRCh38
rs11537782 Chr.X: 15269821-15269821 on Build GRCh38
rs757030999 Chr.X: 15269878-15269878 on Build GRCh38
rs193123788 Chr.X: 15269916-15269916 on Build GRCh38
rs748125526 Chr.X: 15269771-15269771 on Build GRCh38
rs773128197 Chr.X: 15269804-15269804 on Build GRCh38
rs779668682 Chr.X: 15269892-15269892 on Build GRCh38
rs761036835 Chr.X: 15269836-15269836 on Build GRCh38
rs777954576 Chr.X: 15269912-15269912 on Build GRCh38
rs747001148 Chr.X: 15269894-15269894 on Build GRCh38
rs778703434 Chr.X: 15269879-15269879 on Build GRCh38
rs151217271 Chr.X: 15269785-15269785 on Build GRCh38
rs1042504001 Chr.X: 15269838-15269838 on Build GRCh38
rs369829883 Chr.X: 15269919-15269919 on Build GRCh38
rs772202884 Chr.X: 15269782-15269782 on Build GRCh38
rs1023842668 Chr.X: 15269730-15269730 on Build GRCh38
rs771116146 Chr.X: 15269815-15269815 on Build GRCh38
rs769733842 Chr.X: 15269843-15269843 on Build GRCh38
rs746031297 Chr.X: 15269835-15269835 on Build GRCh38
rs774753531 Chr.X: 15269818-15269818 on Build GRCh38
rs145215679 Chr.X: 15269834-15269834 on Build GRCh38
rs753266392 Chr.X: 15269859-15269859 on Build GRCh38
rs753548755 Chr.X: 15269737-15269737 on Build GRCh38
rs188977458 Chr.X: 15269884-15269884 on Build GRCh38
rs756928934 Chr.X: 15269743-15269743 on Build GRCh38
rs745646362 Chr.X: 15269773-15269773 on Build GRCh38
rs921791874 Chr.X: 15269856-15269856 on Build GRCh38
rs759885330 Chr.X: 15269833-15269833 on Build GRCh38
rs768449201 Chr.X: 15269904-15269904 on Build GRCh38
rs777089035 Chr.X: 15269731-15269731 on Build GRCh38
rs201982550 Chr.X: 15269891-15269891 on Build GRCh38
rs763278806 Chr.X: 15269807-15269807 on Build GRCh38
rs774557352 Chr.X: 15269920-15269920 on Build GRCh38

View Full Product Details