Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 38813704-38813935 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHX57
Entrez Gene ID
90957
Gene Name
DEAH-box helicase 57
Gene Aliases
DDX57
UniGene
Hs.468226
Gene Chromosome Location
Chr.2: 38797729-38875883 on Build GRCh38
Transcripts
NM_198963.1 XM_011533154.2 XM_011533155.2 XM_011533156.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4094163 c.3667C>G p.P1223A Chr.2: 38813835-38813835 on Build GRCh38
1250063 c.3619G>A p.A1207T Chr.2: 38813883-38813883 on Build GRCh38
1637970 c.3639A>T p.I1213I Chr.2: 38813863-38813863 on Build GRCh38
4916661 c.3639A>G p.I1213M Chr.2: 38813863-38813863 on Build GRCh38
7232512 c.3622G>A p.E1208K Chr.2: 38813880-38813880 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768049813 Chr.2: 38813778-38813778 on Build GRCh38
rs200596889 Chr.2: 38813803-38813803 on Build GRCh38
rs971819607 Chr.2: 38813766-38813766 on Build GRCh38
rs780009363 Chr.2: 38813831-38813831 on Build GRCh38
rs370295178 Chr.2: 38813826-38813826 on Build GRCh38
rs936316057 Chr.2: 38813779-38813779 on Build GRCh38
rs758314882 Chr.2: 38813830-38813830 on Build GRCh38
rs760021695 Chr.2: 38813771-38813771 on Build GRCh38
rs973064080 Chr.2: 38813740-38813740 on Build GRCh38
rs765474041 Chr.2: 38813907-38813907 on Build GRCh38
rs746909137 Chr.2: 38813843-38813843 on Build GRCh38
rs764554764 Chr.2: 38813894-38813894 on Build GRCh38
rs978290786 Chr.2: 38813764-38813764 on Build GRCh38
rs374656593 Chr.2: 38813762-38813762 on Build GRCh38
rs761815445 Chr.2: 38813748-38813748 on Build GRCh38
rs780899834 Chr.2: 38813857-38813857 on Build GRCh38
rs201516645 Chr.2: 38813816-38813816 on Build GRCh38
rs754122218 Chr.2: 38813811-38813811 on Build GRCh38
rs181107316 Chr.2: 38813899-38813899 on Build GRCh38
rs777128438 Chr.2: 38813898-38813898 on Build GRCh38
rs754781303 Chr.2: 38813856-38813856 on Build GRCh38
rs776167951 Chr.2: 38813888-38813888 on Build GRCh38
rs747864012 Chr.2: 38813862-38813862 on Build GRCh38
rs761201191 Chr.2: 38813889-38813889 on Build GRCh38
rs769476598 Chr.2: 38813868-38813868 on Build GRCh38
rs992332742 Chr.2: 38813746-38813746 on Build GRCh38
rs548777494 Chr.2: 38813809-38813809 on Build GRCh38
rs775654916 Chr.2: 38813747-38813747 on Build GRCh38
rs753047368 Chr.2: 38813780-38813780 on Build GRCh38
rs770310943 Chr.2: 38813875-38813875 on Build GRCh38

View Full Product Details