Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 97909750-97910023 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRMO
Entrez Gene ID
51531
Gene Name
tRNA methyltransferase O
Gene Aliases
C9orf156, HSPC219, NAP1
UniGene
Hs.9196
Gene Chromosome Location
Chr.9: 97876321-97922570 on Build GRCh38
Transcripts
NM_016481.4 XM_005252047.3 XM_005252048.4 XM_005252049.4 XM_005252050.4 XM_011518776.2 XM_011518777.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5802386 c.1038_1062del25 p.E346fs*48 Chr.9: 97909964-97909988 on Build GRCh38
1569817 c.1035C>T p.A345A Chr.9: 97909991-97909991 on Build GRCh38
6315156 c.1028C>A p.P343H Chr.9: 97909998-97909998 on Build GRCh38
1643680 c.1046T>G p.L349R Chr.9: 97909980-97909980 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs762718484 Chr.9: 97909990-97909990 on Build GRCh38
rs1041259818 Chr.9: 97909856-97909856 on Build GRCh38
rs370349301 Chr.9: 97909957-97909957 on Build GRCh38
rs750793522 Chr.9: 97909946-97909946 on Build GRCh38
rs911153808 Chr.9: 97909836-97909836 on Build GRCh38
rs146151071 Chr.9: 97909925-97909925 on Build GRCh38
rs757443842 Chr.9: 97909931-97909931 on Build GRCh38
rs780480936 Chr.9: 97909956-97909956 on Build GRCh38
rs376231163 Chr.9: 97909852-97909852 on Build GRCh38
rs554708441 Chr.9: 97909879-97909879 on Build GRCh38
rs768439647 Chr.9: 97909913-97909914 on Build GRCh38
rs561846135 Chr.9: 97909976-97909976 on Build GRCh38
rs186478738 Chr.9: 97909875-97909875 on Build GRCh38
rs78282249 Chr.9: 97909918-97909918 on Build GRCh38
rs769366984 Chr.9: 97909986-97909986 on Build GRCh38
rs779314985 Chr.9: 97909943-97909943 on Build GRCh38
rs906365479 Chr.9: 97909813-97909813 on Build GRCh38
rs1041825262 Chr.9: 97909866-97909866 on Build GRCh38
rs759579139 Chr.9: 97909997-97909997 on Build GRCh38
rs10984311 Chr.9: 97909861-97909861 on Build GRCh38
rs764276680 Chr.9: 97909920-97909920 on Build GRCh38
rs894914041 Chr.9: 97909878-97909878 on Build GRCh38
rs575926256 Chr.9: 97909795-97909795 on Build GRCh38
rs546156779 Chr.9: 97909842-97909842 on Build GRCh38
rs758930936 Chr.9: 97909950-97909950 on Build GRCh38
rs773059335 Chr.9: 97909987-97909987 on Build GRCh38
rs1048275673 Chr.9: 97909834-97909834 on Build GRCh38
rs747685073 Chr.9: 97909890-97909890 on Build GRCh38
rs770775804 Chr.9: 97909991-97909991 on Build GRCh38
rs901047408 Chr.9: 97909860-97909860 on Build GRCh38
rs374688396 Chr.9: 97909960-97909960 on Build GRCh38
rs143112291 Chr.9: 97909777-97909777 on Build GRCh38
rs1034379130 Chr.9: 97909911-97909911 on Build GRCh38
rs942664764 Chr.9: 97909843-97909843 on Build GRCh38
rs368882020 Chr.9: 97909996-97909996 on Build GRCh38

View Full Product Details