Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 138809993-138810260 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTNNA1
Entrez Gene ID
1495
Gene Name
catenin alpha 1
Gene Aliases
CAP102, MDPT2
UniGene
Hs.445981
Gene Chromosome Location
Chr.5: 138753386-138935034 on Build GRCh38
Transcripts
NM_001290307.2 NM_001290309.2 NM_001290310.2 NM_001323982.1 NM_001323983.1 NM_001323984.1 NM_001323985.1 NM_001323986.1 NM_001903.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5867387 c.403G>A p.V135I Chr.5: 138810139-138810139 on Build GRCh38
5402203 c.377G>A p.R126Q Chr.5: 138810113-138810113 on Build GRCh38
7413575 c.415C>A p.L139M Chr.5: 138810151-138810151 on Build GRCh38
1636821 c.455T>C p.V152A Chr.5: 138810191-138810191 on Build GRCh38
6836570 c.400G>C p.A134P Chr.5: 138810136-138810136 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761084210 Chr.5: 138810083-138810083 on Build GRCh38
rs780843815 Chr.5: 138810057-138810057 on Build GRCh38
rs777986786 Chr.5: 138810139-138810139 on Build GRCh38
rs747636658 Chr.5: 138810204-138810204 on Build GRCh38
rs370783100 Chr.5: 138810177-138810177 on Build GRCh38
rs922655280 Chr.5: 138810142-138810142 on Build GRCh38
rs1059021 Chr.5: 138810153-138810153 on Build GRCh38
rs371054484 Chr.5: 138810113-138810113 on Build GRCh38
rs142395231 Chr.5: 138810093-138810093 on Build GRCh38
rs768136600 Chr.5: 138810074-138810074 on Build GRCh38
rs746920603 Chr.5: 138810052-138810052 on Build GRCh38
rs758599826 Chr.5: 138810121-138810121 on Build GRCh38
rs758513148 Chr.5: 138810035-138810035 on Build GRCh38
rs878854470 Chr.5: 138810031-138810031 on Build GRCh38
rs766845544 Chr.5: 138810086-138810086 on Build GRCh38
rs373849684 Chr.5: 138810215-138810215 on Build GRCh38
rs779734367 Chr.5: 138810160-138810160 on Build GRCh38
rs749878552 Chr.5: 138810098-138810098 on Build GRCh38
rs781124226 Chr.5: 138810145-138810145 on Build GRCh38
rs375976404 Chr.5: 138810221-138810221 on Build GRCh38
rs751579059 Chr.5: 138810141-138810141 on Build GRCh38
rs1043413100 Chr.5: 138810097-138810097 on Build GRCh38
rs769342340 Chr.5: 138810067-138810067 on Build GRCh38
rs773859235 Chr.5: 138810199-138810199 on Build GRCh38
rs762579616 Chr.5: 138810144-138810144 on Build GRCh38
rs775070610 Chr.5: 138810069-138810069 on Build GRCh38
rs762477632 Chr.5: 138810070-138810070 on Build GRCh38
rs1008397016 Chr.5: 138810168-138810168 on Build GRCh38
rs777155637 Chr.5: 138810218-138810218 on Build GRCh38
rs765674162 Chr.5: 138810222-138810222 on Build GRCh38
rs374142395 Chr.5: 138810146-138810146 on Build GRCh38
rs777896797 Chr.5: 138810041-138810041 on Build GRCh38
rs775912588 Chr.5: 138810227-138810227 on Build GRCh38
rs773771416 Chr.5: 138810075-138810075 on Build GRCh38
rs768224635 Chr.5: 138810192-138810192 on Build GRCh38
rs377601723 Chr.5: 138810159-138810159 on Build GRCh38
rs961630194 Chr.5: 138810195-138810195 on Build GRCh38
rs1015730449 Chr.5: 138810182-138810182 on Build GRCh38
rs765587517 Chr.5: 138810106-138810106 on Build GRCh38
rs745606892 Chr.5: 138810063-138810063 on Build GRCh38
rs35788568 Chr.5: 138810108-138810108 on Build GRCh38
rs771365826 Chr.5: 138810216-138810216 on Build GRCh38
rs748930609 Chr.5: 138810166-138810166 on Build GRCh38

View Full Product Details