Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 58517305-58517566 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZBTB45
Entrez Gene ID
84878
Gene Name
zinc finger and BTB domain containing 45
Gene Aliases
ZNF499
UniGene
Hs.515662
Gene Chromosome Location
Chr.19: 58513530-58541712 on Build GRCh38
Transcripts
NM_001316978.1 NM_001316979.1 NM_001316980.1 NM_001316981.1 NM_001316982.1 NM_032792.3 XM_006723445.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6825375 c.157G>A p.A53T Chr.19: 58517517-58517517 on Build GRCh38
6825379 c.142C>T p.R48C Chr.19: 58517532-58517532 on Build GRCh38
5655340 c.161G>T p.G54V Chr.19: 58517513-58517513 on Build GRCh38
6825392 c.242G>A p.R81Q Chr.19: 58517432-58517432 on Build GRCh38
6825367 c.365T>C p.I122T Chr.19: 58517309-58517309 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767505067 Chr.19: 58517519-58517519 on Build GRCh38
rs773622195 Chr.19: 58517432-58517432 on Build GRCh38
rs781185189 Chr.19: 58517395-58517395 on Build GRCh38
rs140420062 Chr.19: 58517433-58517433 on Build GRCh38
rs752890464 Chr.19: 58517516-58517516 on Build GRCh38
rs375511225 Chr.19: 58517445-58517445 on Build GRCh38
rs374408386 Chr.19: 58517527-58517527 on Build GRCh38
rs766517472 Chr.19: 58517438-58517438 on Build GRCh38
rs745503214 Chr.19: 58517396-58517396 on Build GRCh38
rs776806117 Chr.19: 58517506-58517506 on Build GRCh38
rs772435683 Chr.19: 58517472-58517473 on Build GRCh38
rs754976801 Chr.19: 58517443-58517443 on Build GRCh38
rs755086224 Chr.19: 58517363-58517363 on Build GRCh38
rs774189285 Chr.19: 58517414-58517414 on Build GRCh38
rs138240031 Chr.19: 58517476-58517476 on Build GRCh38
rs759855226 Chr.19: 58517507-58517507 on Build GRCh38
rs748867996 Chr.19: 58517532-58517532 on Build GRCh38
rs368790106 Chr.19: 58517485-58517485 on Build GRCh38
rs781131614 Chr.19: 58517454-58517454 on Build GRCh38
rs778279178 Chr.19: 58517546-58517546 on Build GRCh38
rs754715522 Chr.19: 58517544-58517544 on Build GRCh38
rs61735231 Chr.19: 58517326-58517326 on Build GRCh38
rs551242625 Chr.19: 58517359-58517359 on Build GRCh38
rs151253109 Chr.19: 58517402-58517402 on Build GRCh38
rs750409579 Chr.19: 58517523-58517523 on Build GRCh38
rs775287061 Chr.19: 58517398-58517398 on Build GRCh38
rs749212507 Chr.19: 58517390-58517390 on Build GRCh38
rs767955813 Chr.19: 58517329-58517329 on Build GRCh38
rs201382595 Chr.19: 58517389-58517389 on Build GRCh38
rs375219934 Chr.19: 58517356-58517356 on Build GRCh38
rs773959139 Chr.19: 58517482-58517482 on Build GRCh38
rs560957575 Chr.19: 58517488-58517488 on Build GRCh38
rs758413709 Chr.19: 58517518-58517518 on Build GRCh38
rs571138122 Chr.19: 58517444-58517444 on Build GRCh38
rs773710421 Chr.19: 58517490-58517490 on Build GRCh38
rs372758478 Chr.19: 58517526-58517526 on Build GRCh38
rs774680990 Chr.19: 58517407-58517407 on Build GRCh38
rs753983075 Chr.19: 58517362-58517362 on Build GRCh38
rs267605745 Chr.19: 58517325-58517325 on Build GRCh38
rs766748535 Chr.19: 58517498-58517498 on Build GRCh38
rs750611018 Chr.19: 58517350-58517350 on Build GRCh38
rs751431802 Chr.19: 58517446-58517446 on Build GRCh38
rs760968278 Chr.19: 58517494-58517494 on Build GRCh38
rs779804604 Chr.19: 58517465-58517465 on Build GRCh38
rs770668950 Chr.19: 58517401-58517401 on Build GRCh38
rs745594799 Chr.19: 58517455-58517455 on Build GRCh38
rs993861704 Chr.19: 58517382-58517382 on Build GRCh38
rs778217208 Chr.19: 58517522-58517523 on Build GRCh38
rs777509694 Chr.19: 58517375-58517375 on Build GRCh38
rs150431450 Chr.19: 58517466-58517466 on Build GRCh38
rs765221803 Chr.19: 58517515-58517515 on Build GRCh38
rs200526639 Chr.19: 58517397-58517397 on Build GRCh38
rs755987569 Chr.19: 58517456-58517456 on Build GRCh38
rs754069403 Chr.19: 58517442-58517442 on Build GRCh38
rs762421543 Chr.19: 58517412-58517412 on Build GRCh38
rs757280552 Chr.19: 58517448-58517448 on Build GRCh38

View Full Product Details