Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 41510915-41511160 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIVEP3
Entrez Gene ID
59269
Gene Name
human immunodeficiency virus type I enhancer binding protein 3
Gene Aliases
KBP-1, KBP1, KRC, SHN3, Schnurri-3, ZAS3, ZNF40C
UniGene
Hs.403972
Gene Chromosome Location
Chr.1: 41506365-42035925 on Build GRCh38
Transcripts
NM_001127714.2 NM_024503.4 XM_011541884.2 XM_017001992.1 XM_017001993.1 XM_017001994.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4690504 c.6622G>A p.G2208R Chr.1: 41511050-41511050 on Build GRCh38
4504294 c.6615C>T p.A2205A Chr.1: 41511057-41511057 on Build GRCh38
4504623 c.6706C>T p.R2236W Chr.1: 41510966-41510966 on Build GRCh38
1254247 c.6512G>A p.R2171Q Chr.1: 41511160-41511160 on Build GRCh38
4790752 c.6681G>T p.G2227G Chr.1: 41510991-41510991 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752891316 Chr.1: 41511042-41511042 on Build GRCh38
rs767304173 Chr.1: 41510979-41510979 on Build GRCh38
rs779849237 Chr.1: 41510996-41510996 on Build GRCh38
rs753786645 Chr.1: 41510989-41510989 on Build GRCh38
rs770075986 Chr.1: 41511078-41511078 on Build GRCh38
rs147749727 Chr.1: 41510950-41510950 on Build GRCh38
rs758179715 Chr.1: 41510993-41510994 on Build GRCh38
rs776423771 Chr.1: 41511128-41511128 on Build GRCh38
rs201095392 Chr.1: 41510966-41510966 on Build GRCh38
rs769169032 Chr.1: 41510948-41510948 on Build GRCh38
rs745480918 Chr.1: 41510946-41510946 on Build GRCh38
rs745533494 Chr.1: 41510993-41510993 on Build GRCh38
rs367663343 Chr.1: 41510940-41510940 on Build GRCh38
rs1043338184 Chr.1: 41511116-41511116 on Build GRCh38
rs772105236 Chr.1: 41511103-41511103 on Build GRCh38
rs144646554 Chr.1: 41510951-41510951 on Build GRCh38
rs757164295 Chr.1: 41511053-41511053 on Build GRCh38
rs144759096 Chr.1: 41510981-41510981 on Build GRCh38
rs751800969 Chr.1: 41510969-41510969 on Build GRCh38
rs761509178 Chr.1: 41511136-41511136 on Build GRCh38
rs114044801 Chr.1: 41511049-41511049 on Build GRCh38
rs774823137 Chr.1: 41511083-41511083 on Build GRCh38
rs759658128 Chr.1: 41511092-41511092 on Build GRCh38
rs920919846 Chr.1: 41511138-41511138 on Build GRCh38
rs141068012 Chr.1: 41511018-41511018 on Build GRCh38
rs371352900 Chr.1: 41511050-41511050 on Build GRCh38
rs151055315 Chr.1: 41510971-41510971 on Build GRCh38
rs144030867 Chr.1: 41511008-41511008 on Build GRCh38
rs756869288 Chr.1: 41510940-41510941 on Build GRCh38
rs141844226 Chr.1: 41510982-41510982 on Build GRCh38
rs778521437 Chr.1: 41510991-41510991 on Build GRCh38
rs780013598 Chr.1: 41510998-41511005 on Build GRCh38
rs765730736 Chr.1: 41510961-41510961 on Build GRCh38
rs367935226 Chr.1: 41511020-41511020 on Build GRCh38
rs200673170 Chr.1: 41511051-41511051 on Build GRCh38
rs752551787 Chr.1: 41510980-41510980 on Build GRCh38
rs143605709 Chr.1: 41511107-41511107 on Build GRCh38
rs373981666 Chr.1: 41510962-41510962 on Build GRCh38
rs770174463 Chr.1: 41511007-41511007 on Build GRCh38
rs531261702 Chr.1: 41511011-41511011 on Build GRCh38
rs770928569 Chr.1: 41511017-41511017 on Build GRCh38
rs988339270 Chr.1: 41511104-41511104 on Build GRCh38
rs750021373 Chr.1: 41511055-41511055 on Build GRCh38
rs137918317 Chr.1: 41510994-41510994 on Build GRCh38
rs531744210 Chr.1: 41511106-41511106 on Build GRCh38
rs200693003 Chr.1: 41510945-41510945 on Build GRCh38
rs758174985 Chr.1: 41511076-41511076 on Build GRCh38
rs771494084 Chr.1: 41511081-41511081 on Build GRCh38
rs767780667 Chr.1: 41511021-41511021 on Build GRCh38
rs763813067 Chr.1: 41511111-41511111 on Build GRCh38
rs770448762 Chr.1: 41510944-41510944 on Build GRCh38
rs778140478 Chr.1: 41511080-41511080 on Build GRCh38
rs148918385 Chr.1: 41510965-41510965 on Build GRCh38
rs376192406 Chr.1: 41511054-41511054 on Build GRCh38
rs141285859 Chr.1: 41511056-41511056 on Build GRCh38
rs760424322 Chr.1: 41511046-41511046 on Build GRCh38
rs143964973 Chr.1: 41511001-41511001 on Build GRCh38
rs146722099 Chr.1: 41511060-41511060 on Build GRCh38
rs1021097103 Chr.1: 41511114-41511114 on Build GRCh38
rs750187257 Chr.1: 41510986-41510991 on Build GRCh38
rs778462958 Chr.1: 41510941-41510942 on Build GRCh38

View Full Product Details