Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 30282622-30282851 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRTAP24-1
Entrez Gene ID
643803
Gene Name
keratin associated protein 24-1
Gene Aliases
KAP24.1
UniGene
Hs.580883
Gene Chromosome Location
Chr.21: 30281309-30282958 on Build GRCh38
Transcripts
NM_001085455.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
389423 c.262C>T p.P88S Chr.21: 30282671-30282671 on Build GRCh38
6035910 c.190T>A p.S64T Chr.21: 30282743-30282743 on Build GRCh38
7374490 c.292C>T p.P98S Chr.21: 30282641-30282641 on Build GRCh38
1307709 c.183C>G p.C61W Chr.21: 30282750-30282750 on Build GRCh38
3841718 c.92C>A p.S31Y Chr.21: 30282841-30282841 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778725879 Chr.21: 30282820-30282820 on Build GRCh38
rs769825591 Chr.21: 30282739-30282739 on Build GRCh38
rs755447373 Chr.21: 30282822-30282822 on Build GRCh38
rs781593176 Chr.21: 30282738-30282738 on Build GRCh38
rs202216998 Chr.21: 30282796-30282796 on Build GRCh38
rs745593220 Chr.21: 30282646-30282647 on Build GRCh38
rs777321895 Chr.21: 30282648-30282648 on Build GRCh38
rs780495923 Chr.21: 30282709-30282709 on Build GRCh38
rs781131552 Chr.21: 30282644-30282646 on Build GRCh38
rs755618808 Chr.21: 30282681-30282681 on Build GRCh38
rs147639255 Chr.21: 30282645-30282645 on Build GRCh38
rs761834241 Chr.21: 30282664-30282664 on Build GRCh38
rs371296443 Chr.21: 30282737-30282737 on Build GRCh38
rs774083954 Chr.21: 30282775-30282775 on Build GRCh38
rs771156819 Chr.21: 30282767-30282767 on Build GRCh38
rs759412793 Chr.21: 30282672-30282672 on Build GRCh38
rs753389573 Chr.21: 30282798-30282798 on Build GRCh38
rs767465697 Chr.21: 30282678-30282678 on Build GRCh38
rs746211774 Chr.21: 30282653-30282653 on Build GRCh38
rs771768549 Chr.21: 30282670-30282670 on Build GRCh38
rs1039120960 Chr.21: 30282812-30282812 on Build GRCh38
rs753490829 Chr.21: 30282701-30282701 on Build GRCh38
rs759357839 Chr.21: 30282787-30282787 on Build GRCh38
rs373952794 Chr.21: 30282802-30282802 on Build GRCh38
rs376916954 Chr.21: 30282680-30282680 on Build GRCh38
rs776790497 Chr.21: 30282662-30282662 on Build GRCh38
rs924395321 Chr.21: 30282728-30282728 on Build GRCh38
rs201723060 Chr.21: 30282729-30282729 on Build GRCh38
rs777458893 Chr.21: 30282698-30282698 on Build GRCh38
rs755743009 Chr.21: 30282647-30282647 on Build GRCh38
rs772369515 Chr.21: 30282655-30282655 on Build GRCh38
rs760217014 Chr.21: 30282791-30282791 on Build GRCh38
rs201007859 Chr.21: 30282790-30282790 on Build GRCh38
rs944568752 Chr.21: 30282801-30282801 on Build GRCh38
rs767410778 Chr.21: 30282789-30282789 on Build GRCh38
rs978434964 Chr.21: 30282741-30282741 on Build GRCh38
rs773480960 Chr.21: 30282748-30282748 on Build GRCh38
rs372052619 Chr.21: 30282669-30282669 on Build GRCh38
rs922051814 Chr.21: 30282813-30282813 on Build GRCh38
rs747625345 Chr.21: 30282646-30282646 on Build GRCh38
rs751361303 Chr.21: 30282671-30282671 on Build GRCh38
rs925863236 Chr.21: 30282753-30282753 on Build GRCh38
rs777585612 Chr.21: 30282757-30282757 on Build GRCh38
rs368383552 Chr.21: 30282661-30282661 on Build GRCh38
rs758828438 Chr.21: 30282707-30282707 on Build GRCh38
rs375145836 Chr.21: 30282821-30282821 on Build GRCh38
rs770031321 Chr.21: 30282665-30282665 on Build GRCh38
rs756908577 Chr.21: 30282807-30282807 on Build GRCh38
rs769866665 Chr.21: 30282815-30282816 on Build GRCh38
rs780691889 Chr.21: 30282657-30282657 on Build GRCh38

View Full Product Details