Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 103891987-103892234 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OBFC1
Entrez Gene ID
79991
Gene Name
oligonucleotide/oligosaccharide binding fold containing 1
Gene Aliases
AAF-44, AAF44, RPA-32, STN1, bA541N10.2
UniGene
Hs.62314
Gene Chromosome Location
Chr.10: 103856811-103918287 on Build GRCh38
Transcripts
NM_024928.4 XM_006717976.3 XM_011540184.2 XM_017016669.1 XM_017016670.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6355023 c.876+2T>A p.? Chr.10: 103892128-103892128 on Build GRCh38
1345617 c.831delA p.G278fs*16 Chr.10: 103892175-103892175 on Build GRCh38
6732116 c.856G>T p.G286C Chr.10: 103892150-103892150 on Build GRCh38
6351997 c.784G>A p.A262T Chr.10: 103892222-103892222 on Build GRCh38
6732115 c.797T>C p.I266T Chr.10: 103892209-103892209 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753088341 Chr.10: 103892089-103892089 on Build GRCh38
rs921483674 Chr.10: 103892022-103892022 on Build GRCh38
rs940088481 Chr.10: 103892098-103892098 on Build GRCh38
rs755811842 Chr.10: 103892128-103892128 on Build GRCh38
rs778068377 Chr.10: 103892111-103892111 on Build GRCh38
rs760499549 Chr.10: 103892183-103892183 on Build GRCh38
rs544871796 Chr.10: 103892095-103892095 on Build GRCh38
rs566313612 Chr.10: 103892067-103892067 on Build GRCh38
rs770781015 Chr.10: 103892107-103892111 on Build GRCh38
rs758931010 Chr.10: 103892103-103892103 on Build GRCh38
rs931335014 Chr.10: 103892032-103892032 on Build GRCh38
rs199709485 Chr.10: 103892167-103892167 on Build GRCh38
rs758586974 Chr.10: 103892102-103892102 on Build GRCh38
rs748826736 Chr.10: 103892159-103892159 on Build GRCh38
rs772038898 Chr.10: 103892170-103892170 on Build GRCh38
rs201076579 Chr.10: 103892182-103892182 on Build GRCh38
rs201612020 Chr.10: 103892195-103892195 on Build GRCh38
rs765781307 Chr.10: 103892088-103892088 on Build GRCh38
rs746057087 Chr.10: 103892106-103892111 on Build GRCh38
rs1036189827 Chr.10: 103892131-103892131 on Build GRCh38
rs771313798 Chr.10: 103892189-103892189 on Build GRCh38
rs142616199 Chr.10: 103892161-103892161 on Build GRCh38
rs779633657 Chr.10: 103892157-103892157 on Build GRCh38
rs950754576 Chr.10: 103892065-103892065 on Build GRCh38
rs759304639 Chr.10: 103892066-103892066 on Build GRCh38
rs777476869 Chr.10: 103892101-103892102 on Build GRCh38
rs143922724 Chr.10: 103892166-103892166 on Build GRCh38
rs1019246020 Chr.10: 103892135-103892135 on Build GRCh38
rs965987401 Chr.10: 103892180-103892180 on Build GRCh38
rs776930204 Chr.10: 103892190-103892190 on Build GRCh38
rs146867381 Chr.10: 103892197-103892197 on Build GRCh38
rs759652688 Chr.10: 103892086-103892086 on Build GRCh38
rs751891205 Chr.10: 103892123-103892123 on Build GRCh38
rs112272027 Chr.10: 103892035-103892035 on Build GRCh38

View Full Product Details