Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 77591382-77591629 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCEL
Entrez Gene ID
8796
Gene Name
sciellin
Gene Aliases
-
UniGene
Hs.534699
Gene Chromosome Location
Chr.13: 77535674-77645263 on Build GRCh38
Transcripts
NM_001160706.1 NM_003843.3 NM_144777.2 XM_005266578.1 XM_006719882.1 XM_006719884.1 XM_006719885.1 XM_011535281.1 XM_011535282.1 XM_011535283.1 XM_011535284.1 XM_011535285.1 XM_011535286.1 XM_011535287.1 XM_011535288.1 XM_011535289.1 XM_011535290.1 XM_011535291.1 XM_017020805.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
948700 c.661G>T p.E221* Chr.13: 77591429-77591429 on Build GRCh38
4793101 c.646C>T p.P216S Chr.13: 77591414-77591414 on Build GRCh38
1171988 c.659C>T p.T220I Chr.13: 77591427-77591427 on Build GRCh38
3469790 c.671G>A p.R224K Chr.13: 77591439-77591439 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs762634885 Chr.13: 77591422-77591422 on Build GRCh38
rs544106923 Chr.13: 77591551-77591551 on Build GRCh38
rs934204465 Chr.13: 77591492-77591492 on Build GRCh38
rs747523311 Chr.13: 77591485-77591485 on Build GRCh38
rs199947209 Chr.13: 77591509-77591509 on Build GRCh38
rs115680772 Chr.13: 77591502-77591502 on Build GRCh38
rs1048100285 Chr.13: 77591525-77591525 on Build GRCh38
rs182930937 Chr.13: 77591459-77591459 on Build GRCh38
rs752577818 Chr.13: 77591418-77591418 on Build GRCh38
rs771505883 Chr.13: 77591494-77591494 on Build GRCh38
rs918267675 Chr.13: 77591487-77591487 on Build GRCh38
rs386772514 Chr.13: 77591501-77591502 on Build GRCh38
rs751530991 Chr.13: 77591437-77591437 on Build GRCh38
rs757124113 Chr.13: 77591443-77591443 on Build GRCh38
rs993385551 Chr.13: 77591514-77591514 on Build GRCh38
rs779307391 Chr.13: 77591560-77591560 on Build GRCh38
rs77395188 Chr.13: 77591502-77591502 on Build GRCh38
rs757478823 Chr.13: 77591559-77591559 on Build GRCh38
rs753893862 Chr.13: 77591500-77591501 on Build GRCh38
rs775599268 Chr.13: 77591507-77591507 on Build GRCh38
rs770733726 Chr.13: 77591417-77591417 on Build GRCh38
rs41288254 Chr.13: 77591590-77591590 on Build GRCh38
rs1006450714 Chr.13: 77591564-77591567 on Build GRCh38
rs972377799 Chr.13: 77591482-77591482 on Build GRCh38
rs368847500 Chr.13: 77591452-77591452 on Build GRCh38
rs77524201 Chr.13: 77591501-77591501 on Build GRCh38
rs887658907 Chr.13: 77591532-77591532 on Build GRCh38
rs942873981 Chr.13: 77591567-77591567 on Build GRCh38
rs962493363 Chr.13: 77591456-77591456 on Build GRCh38
rs758855272 Chr.13: 77591480-77591480 on Build GRCh38
rs1055072276 Chr.13: 77591575-77591575 on Build GRCh38
rs779539242 Chr.13: 77591464-77591464 on Build GRCh38
rs909626384 Chr.13: 77591433-77591433 on Build GRCh38
rs914780718 Chr.13: 77591460-77591460 on Build GRCh38
rs778241207 Chr.13: 77591483-77591483 on Build GRCh38
rs748591189 Chr.13: 77591468-77591468 on Build GRCh38
rs141475107 Chr.13: 77591430-77591430 on Build GRCh38
rs1010307574 Chr.13: 77591596-77591596 on Build GRCh38
rs780697649 Chr.13: 77591444-77591444 on Build GRCh38
rs147154057 Chr.13: 77591501-77591501 on Build GRCh38

View Full Product Details