Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 88198927-88199198 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM161B
Entrez Gene ID
153396
Gene Name
transmembrane protein 161B
Gene Aliases
FLB3342, PRO1313
UniGene
Hs.379972
Gene Chromosome Location
Chr.5: 88189419-88268917 on Build GRCh38
Transcripts
NM_001289007.1 NM_001289008.1 NM_153354.4 NR_110266.1 XM_006714553.1 XM_006714554.3 XM_006714555.3 XM_006714556.3 XM_011543201.2 XM_011543202.2 XM_011543203.2 XM_011543204.2 XM_017009093.1 XM_017009094.1 XM_017009095.1 XM_017009096.1 XM_017009097.1 XM_017009098.1 XM_017009099.1 XM_017009100.1 XM_017009101.1 XM_017009102.1 XM_017009103.1 XM_017009104.1 XM_017009105.1 XM_017009106.1 XM_017009107.1 XM_017009108.1 XM_017009109.1 XM_017009110.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
738988 c.1072G>A p.V358I Chr.5: 88198993-88198993 on Build GRCh38
450158 c.951G>T p.W317C Chr.5: 88199114-88199114 on Build GRCh38
5672900 c.1083G>T p.Q361H Chr.5: 88198982-88198982 on Build GRCh38
6302239 c.1000C>A p.L334M Chr.5: 88199065-88199065 on Build GRCh38
4166983 c.983C>T p.A328V Chr.5: 88199082-88199082 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs946370799 Chr.5: 88199099-88199099 on Build GRCh38
rs780749535 Chr.5: 88199065-88199065 on Build GRCh38
rs756671409 Chr.5: 88199039-88199039 on Build GRCh38
rs895791506 Chr.5: 88199081-88199081 on Build GRCh38
rs747761653 Chr.5: 88199074-88199074 on Build GRCh38
rs761968910 Chr.5: 88199113-88199113 on Build GRCh38
rs766982004 Chr.5: 88199130-88199130 on Build GRCh38
rs375033902 Chr.5: 88198995-88198995 on Build GRCh38
rs576550087 Chr.5: 88199163-88199163 on Build GRCh38
rs767773974 Chr.5: 88199027-88199027 on Build GRCh38
rs148507249 Chr.5: 88199111-88199111 on Build GRCh38
rs763072448 Chr.5: 88199105-88199105 on Build GRCh38
rs761362064 Chr.5: 88199038-88199038 on Build GRCh38
rs770823544 Chr.5: 88199093-88199093 on Build GRCh38
rs754446018 Chr.5: 88199140-88199140 on Build GRCh38
rs746854349 Chr.5: 88199089-88199089 on Build GRCh38
rs775524809 Chr.5: 88199002-88199002 on Build GRCh38
rs554989470 Chr.5: 88199114-88199114 on Build GRCh38
rs35190770 Chr.5: 88199137-88199137 on Build GRCh38
rs999073989 Chr.5: 88199116-88199116 on Build GRCh38
rs115483226 Chr.5: 88198963-88198963 on Build GRCh38
rs778318163 Chr.5: 88199142-88199142 on Build GRCh38
rs750916496 Chr.5: 88199034-88199034 on Build GRCh38
rs765252105 Chr.5: 88198999-88198999 on Build GRCh38
rs572545951 Chr.5: 88199084-88199084 on Build GRCh38
rs770768628 Chr.5: 88199168-88199168 on Build GRCh38
rs759310312 Chr.5: 88199010-88199010 on Build GRCh38
rs201032572 Chr.5: 88198994-88198994 on Build GRCh38
rs763059487 Chr.5: 88199004-88199004 on Build GRCh38
rs747787451 Chr.5: 88199143-88199143 on Build GRCh38
rs754507674 Chr.5: 88199071-88199071 on Build GRCh38
rs368515045 Chr.5: 88199167-88199167 on Build GRCh38
rs200425900 Chr.5: 88199009-88199009 on Build GRCh38
rs939081884 Chr.5: 88199054-88199054 on Build GRCh38
rs866880439 Chr.5: 88199082-88199082 on Build GRCh38
rs368609687 Chr.5: 88199173-88199173 on Build GRCh38
rs143002306 Chr.5: 88199069-88199069 on Build GRCh38
rs750789831 Chr.5: 88199117-88199117 on Build GRCh38
rs778572006 Chr.5: 88199073-88199073 on Build GRCh38
rs376651198 Chr.5: 88199121-88199121 on Build GRCh38
rs144701087 Chr.5: 88199088-88199088 on Build GRCh38
rs752871590 Chr.5: 88199131-88199131 on Build GRCh38
rs760637396 Chr.5: 88198997-88198997 on Build GRCh38
rs766351143 Chr.5: 88198983-88198983 on Build GRCh38
rs756368846 Chr.5: 88199132-88199132 on Build GRCh38
rs764371176 Chr.5: 88199005-88199005 on Build GRCh38
rs937612952 Chr.5: 88199060-88199060 on Build GRCh38
rs113702172 Chr.5: 88199172-88199172 on Build GRCh38
rs767287137 Chr.5: 88199110-88199110 on Build GRCh38
rs776615655 Chr.5: 88199102-88199102 on Build GRCh38
rs149845262 Chr.5: 88199108-88199108 on Build GRCh38
rs757965279 Chr.5: 88199153-88199153 on Build GRCh38

View Full Product Details