Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 62321543-62321768 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USP15
Entrez Gene ID
9958
Gene Name
ubiquitin specific peptidase 15
Gene Aliases
UNPH-2, UNPH4
UniGene
Hs.434951
Gene Chromosome Location
Chr.12: 62260340-62409721 on Build GRCh38
Transcripts
NM_001252078.1 NM_001252079.1 NM_006313.2 XM_006719718.3 XM_017020284.1 XM_017020285.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6376889 c.584A>G p.D195G Chr.12: 62321572-62321572 on Build GRCh38
3968371 c.576T>C p.N192N Chr.12: 62321564-62321564 on Build GRCh38
6328745 c.601G>T p.A201S Chr.12: 62321589-62321589 on Build GRCh38
6137621 c.565G>C p.E189Q Chr.12: 62321553-62321553 on Build GRCh38
3968370 c.576T>C p.N192N Chr.12: 62321564-62321564 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749483877 Chr.12: 62321631-62321632 on Build GRCh38
rs1004297473 Chr.12: 62321683-62321683 on Build GRCh38
rs1007265669 Chr.12: 62321742-62321742 on Build GRCh38
rs1056111595 Chr.12: 62321576-62321576 on Build GRCh38
rs779727625 Chr.12: 62321629-62321629 on Build GRCh38
rs748207262 Chr.12: 62321669-62321669 on Build GRCh38
rs758172222 Chr.12: 62321602-62321602 on Build GRCh38
rs371431987 Chr.12: 62321636-62321636 on Build GRCh38
rs1051388280 Chr.12: 62321732-62321732 on Build GRCh38
rs758261611 Chr.12: 62321651-62321652 on Build GRCh38
rs750928435 Chr.12: 62321734-62321734 on Build GRCh38
rs201982455 Chr.12: 62321740-62321740 on Build GRCh38
rs750367612 Chr.12: 62321575-62321575 on Build GRCh38
rs929213745 Chr.12: 62321663-62321663 on Build GRCh38
rs111666891 Chr.12: 62321632-62321636 on Build GRCh38
rs199512210 Chr.12: 62321633-62321633 on Build GRCh38
rs571035504 Chr.12: 62321687-62321687 on Build GRCh38
rs372677866 Chr.12: 62321633-62321637 on Build GRCh38
rs779972422 Chr.12: 62321670-62321671 on Build GRCh38
rs372580402 Chr.12: 62321644-62321644 on Build GRCh38
rs781453358 Chr.12: 62321655-62321655 on Build GRCh38

View Full Product Details