Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 36891948-36892189 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF345
Entrez Gene ID
25850
Gene Name
zinc finger protein 345
Gene Aliases
HZF10
UniGene
Hs.362324
Gene Chromosome Location
Chr.19: 36850350-36893218 on Build GRCh38
Transcripts
NR_038362.1 NM_001037232.3 NM_001171979.1 XM_005258876.4 XM_011526933.2

Gene Symbol
ZNF829
Entrez Gene ID
374899
Gene Name
zinc finger protein 829
Gene Aliases
-
UniGene
Hs.720312
Gene Chromosome Location
Chr.19: 36888124-36916387 on Build GRCh38
Transcripts
NR_038362.1 NM_001037232.3 NM_001171979.1 XM_005258876.4 XM_011526933.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4077476 c.625A>G p.K209E Chr.19: 36892166-36892166 on Build GRCh38
1612127 c.829T>A p.F277I Chr.19: 36891962-36891962 on Build GRCh38
6849306 c.731A>T p.E244V Chr.19: 36892060-36892060 on Build GRCh38
4077472 c.722A>C p.E241A Chr.19: 36892069-36892069 on Build GRCh38
4077474 c.660T>C p.A220A Chr.19: 36892131-36892131 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770302636 Chr.19: 36892080-36892080 on Build GRCh38
rs532812185 Chr.19: 36892104-36892104 on Build GRCh38
rs752311498 Chr.19: 36892161-36892161 on Build GRCh38
rs376947168 Chr.19: 36892121-36892121 on Build GRCh38
rs774917493 Chr.19: 36892137-36892137 on Build GRCh38
rs753508299 Chr.19: 36892075-36892075 on Build GRCh38
rs760218666 Chr.19: 36892072-36892072 on Build GRCh38
rs930603104 Chr.19: 36892155-36892155 on Build GRCh38
rs780322855 Chr.19: 36892000-36892000 on Build GRCh38
rs184220904 Chr.19: 36891987-36891987 on Build GRCh38
rs562533250 Chr.19: 36892074-36892074 on Build GRCh38
rs763457141 Chr.19: 36892085-36892085 on Build GRCh38
rs749124125 Chr.19: 36892009-36892009 on Build GRCh38
rs376364399 Chr.19: 36892093-36892093 on Build GRCh38
rs764501501 Chr.19: 36892106-36892106 on Build GRCh38
rs990622782 Chr.19: 36892096-36892096 on Build GRCh38
rs919148631 Chr.19: 36892138-36892138 on Build GRCh38
rs781717744 Chr.19: 36892156-36892156 on Build GRCh38
rs370651502 Chr.19: 36892037-36892037 on Build GRCh38
rs970447971 Chr.19: 36892141-36892141 on Build GRCh38
rs757634184 Chr.19: 36892125-36892125 on Build GRCh38
rs774296155 Chr.19: 36892024-36892024 on Build GRCh38
rs772538159 Chr.19: 36892066-36892066 on Build GRCh38
rs543853566 Chr.19: 36892012-36892012 on Build GRCh38
rs370786414 Chr.19: 36892033-36892033 on Build GRCh38

View Full Product Details