Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81996505-81996772 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASPSCR1
Entrez Gene ID
79058
Gene Name
ASPSCR1, UBX domain containing tether for SLC2A4
Gene Aliases
ASPCR1, ASPL, ASPS, RCC17, TUG, UBXD9, UBXN9
UniGene
Hs.298351
Gene Chromosome Location
Chr.17: 81977550-82018929 on Build GRCh38
Transcripts
NM_001251888.1 NM_024083.3 NR_045351.1 XM_011523601.2 XM_011523602.2 XM_017025036.1 XM_017025037.1 XM_017025038.1 XM_017025039.1 XM_017025040.1 XM_017025041.1 XM_017025042.1 XM_017025043.1 XM_017025044.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5967577 c.771_772insC p.T259fs*10 Chr.17: 81996684-81996685 on Build GRCh38
4506869 c.500C>T p.P167L Chr.17: 81996644-81996644 on Build GRCh38
4429417 c.506C>T p.S169L Chr.17: 81996650-81996650 on Build GRCh38
3796235 c.662C>T p.S221L Chr.17: 81996575-81996575 on Build GRCh38
6410770 c.394C>T p.R132C Chr.17: 81996538-81996538 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757483369 Chr.17: 81996669-81996669 on Build GRCh38
rs770237371 Chr.17: 81996616-81996616 on Build GRCh38
rs367747020 Chr.17: 81996566-81996566 on Build GRCh38
rs753874676 Chr.17: 81996582-81996582 on Build GRCh38
rs186852466 Chr.17: 81996564-81996564 on Build GRCh38
rs998068069 Chr.17: 81996675-81996675 on Build GRCh38
rs752796536 Chr.17: 81996655-81996655 on Build GRCh38
rs61738267 Chr.17: 81996536-81996536 on Build GRCh38
rs748347591 Chr.17: 81996540-81996540 on Build GRCh38
rs372300609 Chr.17: 81996553-81996553 on Build GRCh38
rs781535767 Chr.17: 81996607-81996607 on Build GRCh38
rs775471779 Chr.17: 81996731-81996731 on Build GRCh38
rs778014797 Chr.17: 81996593-81996593 on Build GRCh38
rs781581806 Chr.17: 81996674-81996674 on Build GRCh38
rs773070452 Chr.17: 81996634-81996634 on Build GRCh38
rs752748823 Chr.17: 81996588-81996588 on Build GRCh38
rs368957042 Chr.17: 81996544-81996544 on Build GRCh38
rs764101723 Chr.17: 81996746-81996746 on Build GRCh38
rs764296507 Chr.17: 81996658-81996658 on Build GRCh38
rs761488078 Chr.17: 81996563-81996563 on Build GRCh38
rs765187547 Chr.17: 81996728-81996728 on Build GRCh38
rs903138359 Chr.17: 81996694-81996694 on Build GRCh38
rs919392793 Chr.17: 81996570-81996570 on Build GRCh38
rs372128517 Chr.17: 81996654-81996654 on Build GRCh38
rs1033286860 Chr.17: 81996739-81996739 on Build GRCh38
rs201786107 Chr.17: 81996643-81996643 on Build GRCh38
rs375727115 Chr.17: 81996554-81996554 on Build GRCh38
rs747342381 Chr.17: 81996595-81996595 on Build GRCh38
rs901466624 Chr.17: 81996712-81996712 on Build GRCh38
rs148483554 Chr.17: 81996625-81996625 on Build GRCh38
rs747660580 Chr.17: 81996689-81996689 on Build GRCh38
rs778982611 Chr.17: 81996584-81996584 on Build GRCh38
rs552665655 Chr.17: 81996537-81996537 on Build GRCh38
rs142841603 Chr.17: 81996539-81996539 on Build GRCh38
rs758563653 Chr.17: 81996589-81996589 on Build GRCh38
rs759475392 Chr.17: 81996651-81996651 on Build GRCh38
rs755155984 Chr.17: 81996538-81996538 on Build GRCh38
rs754887850 Chr.17: 81996583-81996583 on Build GRCh38
rs371537189 Chr.17: 81996693-81996693 on Build GRCh38
rs199934562 Chr.17: 81996596-81996596 on Build GRCh38
rs746326324 Chr.17: 81996615-81996615 on Build GRCh38
rs147391651 Chr.17: 81996543-81996543 on Build GRCh38
rs756426183 Chr.17: 81996558-81996558 on Build GRCh38
rs756622860 Chr.17: 81996687-81996687 on Build GRCh38
rs754464527 Chr.17: 81996668-81996668 on Build GRCh38
rs773864878 Chr.17: 81996619-81996619 on Build GRCh38
rs780467570 Chr.17: 81996688-81996688 on Build GRCh38
rs201380867 Chr.17: 81996624-81996624 on Build GRCh38
rs111633399 Chr.17: 81996557-81996557 on Build GRCh38
rs1004550803 Chr.17: 81996528-81996528 on Build GRCh38
rs1048922636 Chr.17: 81996656-81996656 on Build GRCh38
rs150829995 Chr.17: 81996732-81996732 on Build GRCh38
rs771769700 Chr.17: 81996690-81996690 on Build GRCh38
rs772283247 Chr.17: 81996541-81996541 on Build GRCh38
rs766143738 Chr.17: 81996636-81996636 on Build GRCh38
rs760536415 Chr.17: 81996635-81996635 on Build GRCh38
rs8074498 Chr.17: 81996668-81996668 on Build GRCh38
rs751641748 Chr.17: 81996751-81996751 on Build GRCh38
rs766418016 Chr.17: 81996575-81996575 on Build GRCh38
rs776435936 Chr.17: 81996707-81996707 on Build GRCh38
rs759207030 Chr.17: 81996668-81996669 on Build GRCh38
rs759294150 Chr.17: 81996722-81996722 on Build GRCh38

View Full Product Details