Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 49758132-49758403 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMBIM6
Entrez Gene ID
7009
Gene Name
transmembrane BAX inhibitor motif containing 6
Gene Aliases
BAXI1, BI-1, TEGT
UniGene
Hs.743965
Gene Chromosome Location
Chr.12: 49741510-49764934 on Build GRCh38
Transcripts
NM_001098576.1 NM_003217.2 XM_005269126.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5841162 c.293G>A p.G98D Chr.12: 49758233-49758233 on Build GRCh38
3792669 c.299G>A p.G100D Chr.12: 49758239-49758239 on Build GRCh38
4754939 c.504C>A p.N168K Chr.12: 49758270-49758270 on Build GRCh38
6072811 c.300C>T p.G100G Chr.12: 49758240-49758240 on Build GRCh38
223547 c.475C>T p.P159S Chr.12: 49758241-49758241 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753987547 Chr.12: 49758283-49758284 on Build GRCh38
rs747802307 Chr.12: 49758353-49758353 on Build GRCh38
rs764411754 Chr.12: 49758179-49758179 on Build GRCh38
rs554243195 Chr.12: 49758196-49758196 on Build GRCh38
rs768100104 Chr.12: 49758227-49758227 on Build GRCh38
rs146864343 Chr.12: 49758200-49758201 on Build GRCh38
rs757619638 Chr.12: 49758188-49758188 on Build GRCh38
rs2720305 Chr.12: 49758194-49758194 on Build GRCh38
rs202122646 Chr.12: 49758286-49758286 on Build GRCh38
rs74087193 Chr.12: 49758339-49758339 on Build GRCh38
rs778491507 Chr.12: 49758234-49758234 on Build GRCh38
rs754552332 Chr.12: 49758346-49758346 on Build GRCh38
rs895936299 Chr.12: 49758327-49758327 on Build GRCh38
rs113339984 Chr.12: 49758224-49758224 on Build GRCh38
rs968784254 Chr.12: 49758355-49758355 on Build GRCh38
rs61941194 Chr.12: 49758272-49758272 on Build GRCh38
rs764219819 Chr.12: 49758299-49758299 on Build GRCh38
rs901086043 Chr.12: 49758192-49758192 on Build GRCh38
rs200243571 Chr.12: 49758264-49758264 on Build GRCh38
rs73309010 Chr.12: 49758302-49758302 on Build GRCh38
rs780189307 Chr.12: 49758326-49758326 on Build GRCh38
rs775081977 Chr.12: 49758158-49758158 on Build GRCh38
rs776643784 Chr.12: 49758296-49758296 on Build GRCh38
rs369566832 Chr.12: 49758184-49758184 on Build GRCh38
rs771775102 Chr.12: 49758360-49758360 on Build GRCh38
rs773034667 Chr.12: 49758271-49758271 on Build GRCh38
rs748806000 Chr.12: 49758217-49758217 on Build GRCh38
rs750818858 Chr.12: 49758321-49758321 on Build GRCh38
rs754333146 Chr.12: 49758340-49758340 on Build GRCh38
rs756638019 Chr.12: 49758207-49758207 on Build GRCh38
rs149187426 Chr.12: 49758313-49758313 on Build GRCh38
rs370854195 Chr.12: 49758338-49758338 on Build GRCh38
rs924005294 Chr.12: 49758378-49758378 on Build GRCh38
rs111904509 Chr.12: 49758276-49758276 on Build GRCh38
rs369872998 Chr.12: 49758250-49758250 on Build GRCh38
rs771801001 Chr.12: 49758269-49758269 on Build GRCh38
rs191728675 Chr.12: 49758288-49758288 on Build GRCh38
rs1048201554 Chr.12: 49758291-49758291 on Build GRCh38
rs1035305781 Chr.12: 49758309-49758309 on Build GRCh38
rs767086198 Chr.12: 49758352-49758352 on Build GRCh38
rs201212681 Chr.12: 49758216-49758216 on Build GRCh38
rs373091744 Chr.12: 49758193-49758193 on Build GRCh38
rs372405919 Chr.12: 49758208-49758208 on Build GRCh38
rs777246571 Chr.12: 49758377-49758377 on Build GRCh38
rs201337311 Chr.12: 49758314-49758314 on Build GRCh38
rs888240648 Chr.12: 49758247-49758247 on Build GRCh38
rs75489714 Chr.12: 49758298-49758298 on Build GRCh38

View Full Product Details