Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 36367763-36368012 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZFP14
Entrez Gene ID
57677
Gene Name
ZFP14 zinc finger protein
Gene Aliases
ZNF531
UniGene
Hs.35524
Gene Chromosome Location
Chr.19: 36334453-36379203 on Build GRCh38
Transcripts
NM_001297619.1 NM_020917.2 XM_017027038.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3532913 c.7C>T p.H3Y Chr.19: 36367886-36367886 on Build GRCh38
4168753 c.4G>A p.A2T Chr.19: 36367889-36367889 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1013790166 Chr.19: 36367991-36367991 on Build GRCh38
rs777216294 Chr.19: 36367956-36367956 on Build GRCh38
rs200761980 Chr.19: 36367857-36367857 on Build GRCh38
rs760161378 Chr.19: 36367848-36367848 on Build GRCh38
rs763196723 Chr.19: 36367852-36367852 on Build GRCh38
rs767420598 Chr.19: 36367885-36367885 on Build GRCh38
rs895561132 Chr.19: 36367957-36367957 on Build GRCh38
rs1043085483 Chr.19: 36367796-36367796 on Build GRCh38
rs775530038 Chr.19: 36367836-36367836 on Build GRCh38
rs373682984 Chr.19: 36367906-36367906 on Build GRCh38
rs565469311 Chr.19: 36367940-36367940 on Build GRCh38
rs878937821 Chr.19: 36367948-36367948 on Build GRCh38
rs1034141997 Chr.19: 36367954-36367954 on Build GRCh38
rs755520997 Chr.19: 36367917-36367917 on Build GRCh38
rs573328652 Chr.19: 36367856-36367856 on Build GRCh38
rs540841056 Chr.19: 36367955-36367955 on Build GRCh38
rs761726964 Chr.19: 36367870-36367870 on Build GRCh38

View Full Product Details