Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 118395044-118395303 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR44
Entrez Gene ID
54521
Gene Name
WD repeat domain 44
Gene Aliases
RAB11BP, RPH11
UniGene
Hs.98510
Gene Chromosome Location
Chr.X: 118345997-118449961 on Build GRCh38
Transcripts
NM_001184965.1 NM_001184966.1 NM_019045.4 XM_011531353.2 XM_017029600.1

SNP Detail

SNP ID Chromosome Location
rs192688372 Chr.X: 118395181-118395181 on Build GRCh38
rs887709737 Chr.X: 118395261-118395261 on Build GRCh38
rs750053318 Chr.X: 118395251-118395251 on Build GRCh38
rs369804549 Chr.X: 118395147-118395147 on Build GRCh38
rs894957106 Chr.X: 118395113-118395113 on Build GRCh38
rs1012049654 Chr.X: 118395115-118395115 on Build GRCh38
rs1049423287 Chr.X: 118395194-118395194 on Build GRCh38
rs760374300 Chr.X: 118395279-118395279 on Build GRCh38
rs900189476 Chr.X: 118395110-118395110 on Build GRCh38
rs1002819305 Chr.X: 118395183-118395183 on Build GRCh38
rs931685356 Chr.X: 118395186-118395186 on Build GRCh38
rs970656727 Chr.X: 118395160-118395160 on Build GRCh38
rs1015559175 Chr.X: 118395276-118395276 on Build GRCh38
rs766994737 Chr.X: 118395241-118395241 on Build GRCh38

View Full Product Details