Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2232872-2233137 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
E4F1
Entrez Gene ID
1877
Gene Name
E4F transcription factor 1
Gene Aliases
E4F
UniGene
Hs.513268
Gene Chromosome Location
Chr.16: 2223488-2235742 on Build GRCh38
Transcripts
NM_001288776.1 NM_001288778.1 NM_004424.4 XM_005255155.1 XM_005255156.1 XM_006720858.1 XM_011522402.1 XM_011522403.1 XM_017023010.1 XM_017023011.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7427241 c.897A>C p.G299G Chr.16: 2233024-2233024 on Build GRCh38
4679996 c.1008C>T p.H336H Chr.16: 2233135-2233135 on Build GRCh38
7260075 c.874G>A p.A292T Chr.16: 2232899-2232899 on Build GRCh38
5590711 c.941C>T p.P314L Chr.16: 2233068-2233068 on Build GRCh38
6651085 c.991G>A p.V331I Chr.16: 2233118-2233118 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs963203854 Chr.16: 2233054-2233054 on Build GRCh38
rs200055937 Chr.16: 2233108-2233108 on Build GRCh38
rs752735909 Chr.16: 2232913-2232913 on Build GRCh38
rs774104454 Chr.16: 2232963-2232963 on Build GRCh38
rs377655467 Chr.16: 2232895-2232895 on Build GRCh38
rs772665318 Chr.16: 2232945-2232945 on Build GRCh38
rs762313154 Chr.16: 2232978-2232979 on Build GRCh38
rs143221732 Chr.16: 2233039-2233040 on Build GRCh38
rs779313063 Chr.16: 2232937-2232937 on Build GRCh38
rs774016516 Chr.16: 2233056-2233056 on Build GRCh38
rs576565106 Chr.16: 2233048-2233048 on Build GRCh38
rs766785583 Chr.16: 2233064-2233064 on Build GRCh38
rs760682407 Chr.16: 2232994-2232994 on Build GRCh38
rs767396342 Chr.16: 2232981-2232981 on Build GRCh38
rs200965712 Chr.16: 2233053-2233053 on Build GRCh38
rs751372530 Chr.16: 2232903-2232903 on Build GRCh38
rs200987209 Chr.16: 2232941-2232941 on Build GRCh38
rs772755097 Chr.16: 2232891-2232891 on Build GRCh38
rs745605953 Chr.16: 2233063-2233063 on Build GRCh38
rs201773481 Chr.16: 2233047-2233047 on Build GRCh38
rs369274242 Chr.16: 2233117-2233117 on Build GRCh38
rs370872113 Chr.16: 2232908-2232908 on Build GRCh38
rs752646018 Chr.16: 2232985-2232985 on Build GRCh38
rs780638902 Chr.16: 2233034-2233034 on Build GRCh38
rs992690256 Chr.16: 2233094-2233094 on Build GRCh38
rs182042349 Chr.16: 2232921-2232921 on Build GRCh38
rs199627791 Chr.16: 2232902-2232902 on Build GRCh38
rs762585673 Chr.16: 2232899-2232899 on Build GRCh38
rs766832827 Chr.16: 2233116-2233116 on Build GRCh38
rs369317092 Chr.16: 2233118-2233118 on Build GRCh38
rs747601217 Chr.16: 2232952-2232952 on Build GRCh38
rs780726805 Chr.16: 2232951-2232951 on Build GRCh38
rs919031007 Chr.16: 2233092-2233092 on Build GRCh38
rs777310936 Chr.16: 2233041-2233041 on Build GRCh38
rs373679522 Chr.16: 2232955-2232955 on Build GRCh38
rs986016154 Chr.16: 2233046-2233046 on Build GRCh38
rs754240768 Chr.16: 2232969-2232970 on Build GRCh38
rs371870350 Chr.16: 2233084-2233084 on Build GRCh38
rs543017512 Chr.16: 2233036-2233036 on Build GRCh38
rs760648312 Chr.16: 2233075-2233075 on Build GRCh38
rs996242615 Chr.16: 2232964-2232964 on Build GRCh38
rs775330857 Chr.16: 2233067-2233067 on Build GRCh38
rs759361650 Chr.16: 2232968-2232968 on Build GRCh38
rs756210915 Chr.16: 2232915-2232915 on Build GRCh38
rs765497258 Chr.16: 2233093-2233093 on Build GRCh38
rs761819706 Chr.16: 2233085-2233085 on Build GRCh38
rs750767205 Chr.16: 2233030-2233030 on Build GRCh38
rs370540960 Chr.16: 2232953-2232953 on Build GRCh38
rs753924202 Chr.16: 2233019-2233019 on Build GRCh38
rs375309480 Chr.16: 2233026-2233026 on Build GRCh38
rs757488406 Chr.16: 2232922-2232922 on Build GRCh38
rs534584392 Chr.16: 2232956-2232956 on Build GRCh38
rs199667792 Chr.16: 2232962-2232962 on Build GRCh38
rs776250184 Chr.16: 2232893-2232893 on Build GRCh38
rs543875333 Chr.16: 2232898-2232898 on Build GRCh38
rs758845282 Chr.16: 2233031-2233031 on Build GRCh38
rs750675436 Chr.16: 2233106-2233106 on Build GRCh38
rs372248982 Chr.16: 2233011-2233011 on Build GRCh38
rs763978552 Chr.16: 2233078-2233078 on Build GRCh38

View Full Product Details