Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 34219468-34219693 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LSM14A
Entrez Gene ID
26065
Gene Name
LSM14A, mRNA processing body assembly factor
Gene Aliases
C19orf13, FAM61A, RAP55, RAP55A
UniGene
Hs.744009
Gene Chromosome Location
Chr.19: 34172428-34229515 on Build GRCh38
Transcripts
NM_001114093.1 NM_015578.2 XM_005258719.4 XM_005258720.4 XM_005258721.4 XM_011526708.2 XM_017026575.1 XM_017026576.1 XM_017026577.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7160942 c.870G>A p.M290I Chr.19: 34219479-34219479 on Build GRCh38
7160944 c.877G>C p.E293Q Chr.19: 34219486-34219486 on Build GRCh38
2751692 c.901G>T p.A301S Chr.19: 34219510-34219510 on Build GRCh38
5079024 c.934_935delAG p.E313fs*3 Chr.19: 34219543-34219544 on Build GRCh38
5828842 c.959T>A p.L320* Chr.19: 34219568-34219568 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780913140 Chr.19: 34219600-34219602 on Build GRCh38
rs772479671 Chr.19: 34219553-34219553 on Build GRCh38
rs764772465 Chr.19: 34219608-34219608 on Build GRCh38
rs760316791 Chr.19: 34219569-34219569 on Build GRCh38
rs149431669 Chr.19: 34219653-34219653 on Build GRCh38
rs200938297 Chr.19: 34219563-34219563 on Build GRCh38
rs372366966 Chr.19: 34219543-34219543 on Build GRCh38
rs756320607 Chr.19: 34219617-34219617 on Build GRCh38
rs957654381 Chr.19: 34219654-34219654 on Build GRCh38
rs201839474 Chr.19: 34219518-34219518 on Build GRCh38
rs186660789 Chr.19: 34219613-34219613 on Build GRCh38
rs538565321 Chr.19: 34219626-34219626 on Build GRCh38
rs759065472 Chr.19: 34219604-34219604 on Build GRCh38
rs1055714135 Chr.19: 34219564-34219564 on Build GRCh38
rs199952711 Chr.19: 34219662-34219662 on Build GRCh38
rs749413113 Chr.19: 34219503-34219503 on Build GRCh38
rs567009190 Chr.19: 34219494-34219494 on Build GRCh38
rs769238519 Chr.19: 34219507-34219507 on Build GRCh38
rs758392181 Chr.19: 34219615-34219615 on Build GRCh38
rs753303749 Chr.19: 34219578-34219578 on Build GRCh38
rs545590684 Chr.19: 34219664-34219664 on Build GRCh38
rs79410625 Chr.19: 34219658-34219658 on Build GRCh38
rs773505019 Chr.19: 34219560-34219560 on Build GRCh38
rs145856929 Chr.19: 34219663-34219663 on Build GRCh38
rs762450044 Chr.19: 34219548-34219548 on Build GRCh38
rs1026862823 Chr.19: 34219521-34219521 on Build GRCh38
rs778520781 Chr.19: 34219648-34219648 on Build GRCh38
rs765948882 Chr.19: 34219574-34219574 on Build GRCh38
rs369519109 Chr.19: 34219622-34219622 on Build GRCh38
rs375060153 Chr.19: 34219598-34219598 on Build GRCh38
rs2297521 Chr.19: 34219627-34219627 on Build GRCh38
rs750956463 Chr.19: 34219580-34219580 on Build GRCh38
rs757059906 Chr.19: 34219581-34219582 on Build GRCh38
rs763894020 Chr.19: 34219616-34219616 on Build GRCh38

View Full Product Details