Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 99452261-99452504 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP5J2-PTCD1
Entrez Gene ID
100526740
Gene Name
ATP5J2-PTCD1 readthrough
Gene Aliases
-
UniGene
Hs.632313
Gene Chromosome Location
Chr.7: 99416739-99466201 on Build GRCh38
Transcripts
NM_001198879.1 NM_001081559.2 NM_001318160.1 NM_001318161.1 NM_001318162.1 NM_006693.3 XM_011515755.2 XM_011515756.2 XM_011515757.2 XM_017011700.1 XM_017011701.1 XM_017011702.1 XM_017011703.1

Gene Symbol
CPSF4
Entrez Gene ID
10898
Gene Name
cleavage and polyadenylation specific factor 4
Gene Aliases
CPSF30, NAR, NEB-1, NEB1
UniGene
Hs.489287
Gene Chromosome Location
Chr.7: 99438920-99457377 on Build GRCh38
Transcripts
NM_001198879.1 NM_001081559.2 NM_001318160.1 NM_001318161.1 NM_001318162.1 NM_006693.3 XM_011515755.2 XM_011515756.2 XM_011515757.2 XM_017011700.1 XM_017011701.1 XM_017011702.1 XM_017011703.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
453661 c.503G>A p.R168Q Chr.7: 99452373-99452373 on Build GRCh38
7226783 c.545C>T p.P182L Chr.7: 99452415-99452415 on Build GRCh38
3951022 c.545C>A p.P182Q Chr.7: 99452415-99452415 on Build GRCh38
3982302 c.546G>A p.P182P Chr.7: 99452416-99452416 on Build GRCh38
3778731 c.508G>C p.E170Q Chr.7: 99452378-99452378 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs756313629 Chr.7: 99452421-99452421 on Build GRCh38
rs970499975 Chr.7: 99452319-99452319 on Build GRCh38
rs750130501 Chr.7: 99452329-99452331 on Build GRCh38
rs17854665 Chr.7: 99452416-99452416 on Build GRCh38
rs775168753 Chr.7: 99452465-99452465 on Build GRCh38
rs746292107 Chr.7: 99452461-99452461 on Build GRCh38
rs773633692 Chr.7: 99452332-99452332 on Build GRCh38
rs760248771 Chr.7: 99452372-99452372 on Build GRCh38
rs755941860 Chr.7: 99452392-99452394 on Build GRCh38
rs916416369 Chr.7: 99452292-99452292 on Build GRCh38
rs759194963 Chr.7: 99452350-99452350 on Build GRCh38
rs1048378923 Chr.7: 99452384-99452384 on Build GRCh38
rs751746470 Chr.7: 99452411-99452411 on Build GRCh38
rs749791067 Chr.7: 99452445-99452445 on Build GRCh38
rs553790186 Chr.7: 99452409-99452409 on Build GRCh38
rs373719941 Chr.7: 99452463-99452463 on Build GRCh38
rs771222701 Chr.7: 99452337-99452337 on Build GRCh38
rs757032878 Chr.7: 99452401-99452401 on Build GRCh38
rs774015501 Chr.7: 99452342-99452342 on Build GRCh38
rs571436369 Chr.7: 99452334-99452334 on Build GRCh38
rs369703870 Chr.7: 99452446-99452446 on Build GRCh38
rs766960857 Chr.7: 99452357-99452357 on Build GRCh38
rs150662458 Chr.7: 99452398-99452398 on Build GRCh38
rs774597125 Chr.7: 99452448-99452448 on Build GRCh38
rs755235208 Chr.7: 99452415-99452415 on Build GRCh38
rs748294203 Chr.7: 99452326-99452326 on Build GRCh38
rs760126489 Chr.7: 99452478-99452478 on Build GRCh38
rs112949768 Chr.7: 99452329-99452329 on Build GRCh38
rs200506342 Chr.7: 99452373-99452373 on Build GRCh38
rs768078821 Chr.7: 99452480-99452480 on Build GRCh38
rs377662921 Chr.7: 99452362-99452362 on Build GRCh38
rs918676494 Chr.7: 99452346-99452346 on Build GRCh38
rs778293972 Chr.7: 99452434-99452434 on Build GRCh38

View Full Product Details