Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 59006399-59006624 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTSZ
Entrez Gene ID
1522
Gene Name
cathepsin Z
Gene Aliases
CTSX
UniGene
Hs.252549
Gene Chromosome Location
Chr.20: 58995187-59007254 on Build GRCh38
Transcripts
NM_001336.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5618904 c.192G>A p.K64K Chr.20: 59006437-59006437 on Build GRCh38
725018 c.165G>A p.E55E Chr.20: 59006464-59006464 on Build GRCh38
2765000 c.214G>C p.D72H Chr.20: 59006415-59006415 on Build GRCh38
7363285 c.214G>A p.D72N Chr.20: 59006415-59006415 on Build GRCh38
6558937 c.203G>A p.W68* Chr.20: 59006426-59006426 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764926142 Chr.20: 59006431-59006431 on Build GRCh38
rs898663425 Chr.20: 59006467-59006467 on Build GRCh38
rs764984182 Chr.20: 59006498-59006498 on Build GRCh38
rs1023427201 Chr.20: 59006533-59006533 on Build GRCh38
rs780701364 Chr.20: 59006600-59006600 on Build GRCh38
rs761643208 Chr.20: 59006495-59006495 on Build GRCh38
rs754533124 Chr.20: 59006513-59006513 on Build GRCh38
rs776414207 Chr.20: 59006422-59006422 on Build GRCh38
rs757883716 Chr.20: 59006433-59006433 on Build GRCh38
rs776534185 Chr.20: 59006486-59006486 on Build GRCh38
rs768200449 Chr.20: 59006519-59006519 on Build GRCh38
rs753235038 Chr.20: 59006529-59006529 on Build GRCh38
rs968302966 Chr.20: 59006544-59006544 on Build GRCh38
rs755559076 Chr.20: 59006457-59006457 on Build GRCh38
rs918868200 Chr.20: 59006547-59006547 on Build GRCh38
rs748592572 Chr.20: 59006461-59006461 on Build GRCh38
rs749046807 Chr.20: 59006452-59006453 on Build GRCh38
rs749496207 Chr.20: 59006473-59006473 on Build GRCh38
rs756589555 Chr.20: 59006536-59006536 on Build GRCh38
rs111756806 Chr.20: 59006439-59006439 on Build GRCh38
rs930209287 Chr.20: 59006554-59006554 on Build GRCh38
rs143686598 Chr.20: 59006507-59006507 on Build GRCh38
rs771018508 Chr.20: 59006478-59006478 on Build GRCh38
rs772838602 Chr.20: 59006500-59006500 on Build GRCh38
rs751089751 Chr.20: 59006510-59006510 on Build GRCh38
rs531348521 Chr.20: 59006546-59006546 on Build GRCh38
rs774331858 Chr.20: 59006475-59006475 on Build GRCh38
rs750064793 Chr.20: 59006432-59006432 on Build GRCh38
rs756802063 Chr.20: 59006569-59006569 on Build GRCh38
rs995581034 Chr.20: 59006475-59006475 on Build GRCh38
rs974236883 Chr.20: 59006543-59006543 on Build GRCh38
rs374624286 Chr.20: 59006503-59006503 on Build GRCh38
rs774512938 Chr.20: 59006479-59006479 on Build GRCh38
rs768646415 Chr.20: 59006482-59006482 on Build GRCh38
rs1056135344 Chr.20: 59006423-59006423 on Build GRCh38
rs1056165806 Chr.20: 59006562-59006562 on Build GRCh38
rs781739366 Chr.20: 59006458-59006458 on Build GRCh38
rs1000664531 Chr.20: 59006499-59006499 on Build GRCh38
rs768543182 Chr.20: 59006457-59006457 on Build GRCh38
rs955483601 Chr.20: 59006511-59006511 on Build GRCh38
rs756536654 Chr.20: 59006462-59006462 on Build GRCh38
rs186380851 Chr.20: 59006454-59006454 on Build GRCh38
rs140246463 Chr.20: 59006442-59006442 on Build GRCh38
rs1023353788 Chr.20: 59006456-59006456 on Build GRCh38
rs74350367 Chr.20: 59006468-59006468 on Build GRCh38
rs201197341 Chr.20: 59006435-59006435 on Build GRCh38

View Full Product Details