Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 66602576-66602849 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EDA2R
Entrez Gene ID
60401
Gene Name
ectodysplasin A2 receptor
Gene Aliases
EDA-A2R, EDAA2R, TNFRSF27, XEDAR
UniGene
Hs.302017
Gene Chromosome Location
Chr.X: 66594384-66639303 on Build GRCh38
Transcripts
NM_001199687.2 NM_001242310.1 NM_001324199.1 NM_001324201.1 NM_001324202.1 NM_001324204.1 NM_001324205.1 NM_001324206.1 NM_021783.4 NR_136726.1 NR_136727.1 XM_011531000.1 XM_011531001.1 XM_011531002.1 XM_011531003.2 XM_011531004.1 XM_011531005.2 XM_011531006.2 XM_011531007.2 XM_017029703.1 XM_017029704.1 XM_017029705.1 XM_017029706.1 XM_017029707.1 XM_017029708.1

SNP Detail

SNP ID Chromosome Location
rs1385698 Chr.X: 66602765-66602765 on Build GRCh38
rs760050240 Chr.X: 66602665-66602665 on Build GRCh38
rs867737802 Chr.X: 66602705-66602705 on Build GRCh38
rs776421145 Chr.X: 66602673-66602673 on Build GRCh38
rs765098590 Chr.X: 66602703-66602703 on Build GRCh38
rs746070590 Chr.X: 66602760-66602760 on Build GRCh38
rs758910551 Chr.X: 66602666-66602666 on Build GRCh38
rs769082256 Chr.X: 66602777-66602777 on Build GRCh38
rs868065042 Chr.X: 66602706-66602706 on Build GRCh38
rs780202146 Chr.X: 66602685-66602686 on Build GRCh38
rs774944548 Chr.X: 66602640-66602640 on Build GRCh38
rs141563563 Chr.X: 66602771-66602771 on Build GRCh38
rs200503724 Chr.X: 66602750-66602750 on Build GRCh38
rs770555125 Chr.X: 66602797-66602797 on Build GRCh38
rs781314413 Chr.X: 66602757-66602757 on Build GRCh38
rs758239780 Chr.X: 66602721-66602721 on Build GRCh38
rs749773022 Chr.X: 66602636-66602636 on Build GRCh38
rs146403451 Chr.X: 66602772-66602772 on Build GRCh38
rs747797955 Chr.X: 66602817-66602817 on Build GRCh38
rs776909447 Chr.X: 66602806-66602807 on Build GRCh38
rs779459293 Chr.X: 66602635-66602635 on Build GRCh38
rs143335692 Chr.X: 66602742-66602742 on Build GRCh38
rs752471149 Chr.X: 66602712-66602712 on Build GRCh38
rs896973552 Chr.X: 66602688-66602688 on Build GRCh38
rs1055399292 Chr.X: 66602741-66602741 on Build GRCh38
rs377309250 Chr.X: 66602806-66602806 on Build GRCh38
rs763888781 Chr.X: 66602729-66602729 on Build GRCh38
rs61761340 Chr.X: 66602637-66602637 on Build GRCh38
rs149166760 Chr.X: 66602675-66602675 on Build GRCh38
rs774658464 Chr.X: 66602795-66602795 on Build GRCh38
rs759765858 Chr.X: 66602815-66602815 on Build GRCh38
rs938423758 Chr.X: 66602658-66602658 on Build GRCh38
rs111474487 Chr.X: 66602745-66602745 on Build GRCh38
rs764786622 Chr.X: 66602752-66602752 on Build GRCh38
rs778572925 Chr.X: 66602799-66602799 on Build GRCh38
rs769528529 Chr.X: 66602818-66602818 on Build GRCh38
rs931111155 Chr.X: 66602696-66602696 on Build GRCh38

View Full Product Details