Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 7994339-7994598 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF3F
Entrez Gene ID
8665
Gene Name
eukaryotic translation initiation factor 3 subunit F
Gene Aliases
EIF3S5, eIF3-p47
UniGene
Hs.516023
Gene Chromosome Location
Chr.11: 7987320-7996171 on Build GRCh38
Transcripts
NM_003754.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3368581 c.654-2A>T p.? Chr.11: 7994424-7994424 on Build GRCh38
7092848 c.699G>A p.T233T Chr.11: 7994471-7994471 on Build GRCh38
690491 c.745+1G>T p.? Chr.11: 7994518-7994518 on Build GRCh38
5787165 c.748C>T p.L250L Chr.11: 7994475-7994475 on Build GRCh38
7092849 c.744G>A p.T248T Chr.11: 7994471-7994471 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141976414 Chr.11: 7994466-7994466 on Build GRCh38
rs113494623 Chr.11: 7994518-7994518 on Build GRCh38
rs776919549 Chr.11: 7994421-7994421 on Build GRCh38
rs750576391 Chr.11: 7994529-7994529 on Build GRCh38
rs575801173 Chr.11: 7994573-7994573 on Build GRCh38
rs145163086 Chr.11: 7994491-7994491 on Build GRCh38
rs756483530 Chr.11: 7994569-7994569 on Build GRCh38
rs926455527 Chr.11: 7994557-7994557 on Build GRCh38
rs572472001 Chr.11: 7994578-7994578 on Build GRCh38
rs7102030 Chr.11: 7994498-7994498 on Build GRCh38
rs770529976 Chr.11: 7994423-7994423 on Build GRCh38
rs747544996 Chr.11: 7994414-7994414 on Build GRCh38
rs758431097 Chr.11: 7994471-7994471 on Build GRCh38
rs765487803 Chr.11: 7994468-7994468 on Build GRCh38
rs200510904 Chr.11: 7994453-7994453 on Build GRCh38
rs1021979682 Chr.11: 7994486-7994486 on Build GRCh38
rs545728244 Chr.11: 7994532-7994532 on Build GRCh38
rs372196261 Chr.11: 7994541-7994541 on Build GRCh38
rs753282970 Chr.11: 7994564-7994564 on Build GRCh38
rs758877237 Chr.11: 7994403-7994403 on Build GRCh38
rs375100719 Chr.11: 7994388-7994388 on Build GRCh38
rs773976539 Chr.11: 7994433-7994433 on Build GRCh38
rs755901524 Chr.11: 7994378-7994378 on Build GRCh38
rs781502960 Chr.11: 7994563-7994563 on Build GRCh38
rs771594375 Chr.11: 7994533-7994533 on Build GRCh38
rs780110207 Chr.11: 7994473-7994473 on Build GRCh38
rs760654379 Chr.11: 7994511-7994511 on Build GRCh38
rs748967109 Chr.11: 7994422-7994422 on Build GRCh38
rs748512155 Chr.11: 7994497-7994497 on Build GRCh38
rs369989218 Chr.11: 7994420-7994420 on Build GRCh38
rs372113064 Chr.11: 7994390-7994390 on Build GRCh38
rs775504098 Chr.11: 7994457-7994457 on Build GRCh38
rs761462188 Chr.11: 7994514-7994514 on Build GRCh38
rs1048061834 Chr.11: 7994374-7994374 on Build GRCh38
rs745523675 Chr.11: 7994501-7994501 on Build GRCh38
rs766443567 Chr.11: 7994540-7994540 on Build GRCh38
rs892704019 Chr.11: 7994450-7994450 on Build GRCh38
rs763804451 Chr.11: 7994383-7994383 on Build GRCh38
rs755489020 Chr.11: 7994543-7994543 on Build GRCh38
rs745984688 Chr.11: 7994396-7994396 on Build GRCh38
rs778672688 Chr.11: 7994499-7994499 on Build GRCh38
rs760545005 Chr.11: 7994463-7994463 on Build GRCh38
rs868644022 Chr.11: 7994458-7994458 on Build GRCh38
rs564459532 Chr.11: 7994489-7994489 on Build GRCh38
rs966351855 Chr.11: 7994411-7994411 on Build GRCh38
rs138812206 Chr.11: 7994492-7994492 on Build GRCh38
rs372549173 Chr.11: 7994490-7994490 on Build GRCh38
rs757476263 Chr.11: 7994393-7994393 on Build GRCh38
rs780489360 Chr.11: 7994405-7994405 on Build GRCh38
rs753640542 Chr.11: 7994465-7994465 on Build GRCh38
rs765047822 Chr.11: 7994525-7994525 on Build GRCh38
rs553827428 Chr.11: 7994363-7994363 on Build GRCh38
rs763932999 Chr.11: 7994464-7994464 on Build GRCh38
rs756781059 Chr.11: 7994568-7994569 on Build GRCh38
rs201665082 Chr.11: 7994509-7994509 on Build GRCh38
rs570574577 Chr.11: 7994513-7994513 on Build GRCh38
rs375398038 Chr.11: 7994395-7994395 on Build GRCh38
rs759072919 Chr.11: 7994440-7994440 on Build GRCh38
rs750665696 Chr.11: 7994469-7994469 on Build GRCh38

View Full Product Details