Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 50548900-50549155 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPATA20
Entrez Gene ID
64847
Gene Name
spermatogenesis associated 20
Gene Aliases
HEL-S-98, SSP411, Tisp78
UniGene
Hs.103147
Gene Chromosome Location
Chr.17: 50547089-50555852 on Build GRCh38
Transcripts
NM_001258372.1 NM_001258373.1 NM_022827.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3519157 c.624C>T p.V208V Chr.17: 50549150-50549150 on Build GRCh38
3519156 c.622G>T p.V208F Chr.17: 50549148-50549148 on Build GRCh38
6035685 c.469C>T p.R157C Chr.17: 50548917-50548917 on Build GRCh38
3519155 c.599C>T p.P200L Chr.17: 50549125-50549125 on Build GRCh38
4501814 c.598C>T p.P200S Chr.17: 50549124-50549124 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757144489 Chr.17: 50549087-50549087 on Build GRCh38
rs750995991 Chr.17: 50549055-50549055 on Build GRCh38
rs923736431 Chr.17: 50549124-50549124 on Build GRCh38
rs747231561 Chr.17: 50549020-50549020 on Build GRCh38
rs752337298 Chr.17: 50549059-50549059 on Build GRCh38
rs916621531 Chr.17: 50549080-50549080 on Build GRCh38
rs752371125 Chr.17: 50548994-50548994 on Build GRCh38
rs375908172 Chr.17: 50548955-50548955 on Build GRCh38
rs991217209 Chr.17: 50549076-50549076 on Build GRCh38
rs770283091 Chr.17: 50549044-50549044 on Build GRCh38
rs8077316 Chr.17: 50549028-50549028 on Build GRCh38
rs376625784 Chr.17: 50549072-50549072 on Build GRCh38
rs372517018 Chr.17: 50549032-50549032 on Build GRCh38
rs764593475 Chr.17: 50548951-50548951 on Build GRCh38
rs140022114 Chr.17: 50549057-50549057 on Build GRCh38
rs141302744 Chr.17: 50548924-50548924 on Build GRCh38
rs779052203 Chr.17: 50549094-50549094 on Build GRCh38
rs376965141 Chr.17: 50549060-50549060 on Build GRCh38
rs140922230 Chr.17: 50549132-50549132 on Build GRCh38
rs777746118 Chr.17: 50548998-50548998 on Build GRCh38
rs200061579 Chr.17: 50549017-50549017 on Build GRCh38
rs749063147 Chr.17: 50548992-50548992 on Build GRCh38
rs201429533 Chr.17: 50549058-50549058 on Build GRCh38
rs778839011 Chr.17: 50549026-50549026 on Build GRCh38
rs777085347 Chr.17: 50549129-50549129 on Build GRCh38
rs754379589 Chr.17: 50548960-50548960 on Build GRCh38
rs982866224 Chr.17: 50549109-50549109 on Build GRCh38
rs8077323 Chr.17: 50549037-50549037 on Build GRCh38
rs765953689 Chr.17: 50548987-50548987 on Build GRCh38
rs751271834 Chr.17: 50548989-50548989 on Build GRCh38
rs893448615 Chr.17: 50548958-50548958 on Build GRCh38
rs766292074 Chr.17: 50548950-50548950 on Build GRCh38
rs372899734 Chr.17: 50548934-50548934 on Build GRCh38
rs201018841 Chr.17: 50549119-50549119 on Build GRCh38
rs201623031 Chr.17: 50549027-50549027 on Build GRCh38
rs764638265 Chr.17: 50548959-50548959 on Build GRCh38
rs369694182 Chr.17: 50548995-50548995 on Build GRCh38
rs774703042 Chr.17: 50548927-50548927 on Build GRCh38
rs753825082 Chr.17: 50549074-50549074 on Build GRCh38
rs770133444 Chr.17: 50549136-50549136 on Build GRCh38
rs757543788 Chr.17: 50549009-50549009 on Build GRCh38
rs150798114 Chr.17: 50549054-50549054 on Build GRCh38
rs549440861 Chr.17: 50548981-50548981 on Build GRCh38
rs749220077 Chr.17: 50549003-50549003 on Build GRCh38
rs780207958 Chr.17: 50549113-50549113 on Build GRCh38
rs866074779 Chr.17: 50548980-50548980 on Build GRCh38
rs370849887 Chr.17: 50549108-50549108 on Build GRCh38
rs1033189592 Chr.17: 50549045-50549045 on Build GRCh38
rs191069312 Chr.17: 50548926-50548926 on Build GRCh38
rs745874128 Chr.17: 50549096-50549096 on Build GRCh38
rs775675215 Chr.17: 50548954-50548954 on Build GRCh38
rs781075277 Chr.17: 50548993-50548993 on Build GRCh38
rs777314435 Chr.17: 50549038-50549038 on Build GRCh38

View Full Product Details