Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 73863099-73863364 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRPL48
Entrez Gene ID
51642
Gene Name
mitochondrial ribosomal protein L48
Gene Aliases
CGI-118, HSPC290, L48MT, MRP-L48
UniGene
Hs.503239
Gene Chromosome Location
Chr.11: 73787872-73864611 on Build GRCh38
Transcripts
NM_001318498.1 NM_001318499.1 NM_001318500.1 NM_016055.5 NR_134658.1 NR_134659.1 NR_134660.1 NR_134661.1 XM_017017919.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
429827 c.518T>A p.I173K Chr.11: 73863215-73863215 on Build GRCh38
5015039 c.518T>C p.I173T Chr.11: 73863215-73863215 on Build GRCh38
4428904 c.536C>T p.P179L Chr.11: 73863233-73863233 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs866351665 Chr.11: 73863220-73863220 on Build GRCh38
rs779431057 Chr.11: 73863279-73863279 on Build GRCh38
rs780546955 Chr.11: 73863185-73863185 on Build GRCh38
rs774243467 Chr.11: 73863300-73863300 on Build GRCh38
rs376288296 Chr.11: 73863120-73863121 on Build GRCh38
rs1005806169 Chr.11: 73863119-73863120 on Build GRCh38
rs543805995 Chr.11: 73863124-73863124 on Build GRCh38
rs201680435 Chr.11: 73863228-73863228 on Build GRCh38
rs757664413 Chr.11: 73863232-73863232 on Build GRCh38
rs375332756 Chr.11: 73863178-73863178 on Build GRCh38
rs755730773 Chr.11: 73863265-73863265 on Build GRCh38
rs899932146 Chr.11: 73863163-73863163 on Build GRCh38
rs759754873 Chr.11: 73863145-73863145 on Build GRCh38
rs373856472 Chr.11: 73863293-73863293 on Build GRCh38
rs866641089 Chr.11: 73863238-73863238 on Build GRCh38
rs955406704 Chr.11: 73863257-73863257 on Build GRCh38
rs561642415 Chr.11: 73863330-73863330 on Build GRCh38
rs868841625 Chr.11: 73863177-73863177 on Build GRCh38
rs11827393 Chr.11: 73863264-73863264 on Build GRCh38
rs375899391 Chr.11: 73863125-73863125 on Build GRCh38
rs778409092 Chr.11: 73863226-73863226 on Build GRCh38
rs748159073 Chr.11: 73863155-73863157 on Build GRCh38
rs1001859284 Chr.11: 73863122-73863122 on Build GRCh38
rs142282360 Chr.11: 73863335-73863335 on Build GRCh38
rs376790843 Chr.11: 73863162-73863162 on Build GRCh38
rs748824144 Chr.11: 73863287-73863287 on Build GRCh38
rs531371956 Chr.11: 73863192-73863192 on Build GRCh38
rs202232453 Chr.11: 73863179-73863179 on Build GRCh38
rs774601912 Chr.11: 73863285-73863285 on Build GRCh38
rs202224445 Chr.11: 73863241-73863241 on Build GRCh38
rs983074794 Chr.11: 73863191-73863191 on Build GRCh38
rs752757627 Chr.11: 73863171-73863171 on Build GRCh38
rs752018643 Chr.11: 73863231-73863231 on Build GRCh38
rs969162531 Chr.11: 73863119-73863119 on Build GRCh38
rs962477550 Chr.11: 73863195-73863195 on Build GRCh38

View Full Product Details