Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 40775702-40775933 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIA
Entrez Gene ID
8190
Gene Name
melanoma inhibitory activity
Gene Aliases
CD-RAP
UniGene
Hs.646364
Gene Chromosome Location
Chr.19: 40775177-40777493 on Build GRCh38
Transcripts
NM_001202553.1 NM_006533.3 NR_037775.1

Gene Symbol
MIA-RAB4B
Entrez Gene ID
100529262
Gene Name
MIA-RAB4B readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.631539
Gene Chromosome Location
Chr.19: 40775537-40796944 on Build GRCh38
Transcripts
NM_001202553.1 NM_006533.3 NR_037775.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3534205 c.156G>A p.Q52Q Chr.19: 40775780-40775780 on Build GRCh38
996824 c.137C>T p.S46F Chr.19: 40775761-40775761 on Build GRCh38
352744 c.186G>A p.L62L Chr.19: 40775810-40775810 on Build GRCh38
6712384 c.250_251insG p.G86fs*21 Chr.19: 40775874-40775875 on Build GRCh38
6465776 c.172G>C p.D58H Chr.19: 40775796-40775796 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758570130 Chr.19: 40775823-40775823 on Build GRCh38
rs149098807 Chr.19: 40775820-40775820 on Build GRCh38
rs755216468 Chr.19: 40775845-40775845 on Build GRCh38
rs775191339 Chr.19: 40775803-40775803 on Build GRCh38
rs771629120 Chr.19: 40775876-40775876 on Build GRCh38
rs758349845 Chr.19: 40775761-40775761 on Build GRCh38
rs751877558 Chr.19: 40775834-40775834 on Build GRCh38
rs371955085 Chr.19: 40775748-40775748 on Build GRCh38
rs748639206 Chr.19: 40775787-40775787 on Build GRCh38
rs755447826 Chr.19: 40775773-40775773 on Build GRCh38
rs778453963 Chr.19: 40775895-40775895 on Build GRCh38
rs78490969 Chr.19: 40775770-40775770 on Build GRCh38
rs771506464 Chr.19: 40775802-40775802 on Build GRCh38
rs368482748 Chr.19: 40775885-40775885 on Build GRCh38
rs778351028 Chr.19: 40775864-40775864 on Build GRCh38
rs200289498 Chr.19: 40775866-40775866 on Build GRCh38
rs756380131 Chr.19: 40775860-40775860 on Build GRCh38
rs2233156 Chr.19: 40775795-40775795 on Build GRCh38
rs764935423 Chr.19: 40775907-40775907 on Build GRCh38
rs900831979 Chr.19: 40775775-40775775 on Build GRCh38
rs781670214 Chr.19: 40775776-40775776 on Build GRCh38
rs746704827 Chr.19: 40775886-40775886 on Build GRCh38
rs748249239 Chr.19: 40775740-40775741 on Build GRCh38
rs1016765119 Chr.19: 40775839-40775839 on Build GRCh38
rs772655092 Chr.19: 40775874-40775875 on Build GRCh38
rs776088164 Chr.19: 40775902-40775902 on Build GRCh38
rs768329055 Chr.19: 40775890-40775890 on Build GRCh38
rs545812317 Chr.19: 40775865-40775865 on Build GRCh38
rs371870072 Chr.19: 40775878-40775878 on Build GRCh38
rs750259359 Chr.19: 40775821-40775821 on Build GRCh38
rs139650429 Chr.19: 40775768-40775769 on Build GRCh38
rs770368729 Chr.19: 40775788-40775788 on Build GRCh38
rs761466075 Chr.19: 40775817-40775817 on Build GRCh38
rs748572393 Chr.19: 40775859-40775859 on Build GRCh38
rs745398952 Chr.19: 40775796-40775796 on Build GRCh38
rs781520113 Chr.19: 40775847-40775847 on Build GRCh38
rs776380625 Chr.19: 40775814-40775814 on Build GRCh38
rs747366993 Chr.19: 40775906-40775907 on Build GRCh38
rs150386826 Chr.19: 40775758-40775758 on Build GRCh38
rs991241625 Chr.19: 40775909-40775909 on Build GRCh38
rs375296959 Chr.19: 40775831-40775831 on Build GRCh38
rs763926172 Chr.19: 40775812-40775812 on Build GRCh38
rs780426282 Chr.19: 40775763-40775763 on Build GRCh38

View Full Product Details